Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 40853392-40853653 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD6
Entrez Gene ID
84674
Gene Name
caspase recruitment domain family member 6
Gene Aliases
CINCIN1
UniGene
Hs.200242
Gene Chromosome Location
Chr.5: 40841190-40855354 on Build GRCh38
Transcripts
NM_032587.3 XM_017009989.1 XM_017009990.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3855278 c.2261G>A p.G754D Chr.5: 40853593-40853593 on Build GRCh38
6513010 c.2179G>A p.V727I Chr.5: 40853511-40853511 on Build GRCh38
5035636 c.2314C>G p.Q772E Chr.5: 40853646-40853646 on Build GRCh38
5036490 c.2187G>C p.E729D Chr.5: 40853519-40853519 on Build GRCh38
389524 c.2107C>G p.Q703E Chr.5: 40853439-40853439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769887763 Chr.5: 40853553-40853553 on Build GRCh38
rs764203570 Chr.5: 40853471-40853471 on Build GRCh38
rs767718486 Chr.5: 40853591-40853591 on Build GRCh38
rs566225791 Chr.5: 40853569-40853569 on Build GRCh38
rs145200542 Chr.5: 40853609-40853609 on Build GRCh38
rs776306921 Chr.5: 40853547-40853547 on Build GRCh38
rs750718581 Chr.5: 40853496-40853496 on Build GRCh38
rs867628058 Chr.5: 40853599-40853599 on Build GRCh38
rs745613212 Chr.5: 40853551-40853551 on Build GRCh38
rs777211768 Chr.5: 40853605-40853605 on Build GRCh38
rs757489251 Chr.5: 40853483-40853483 on Build GRCh38
rs963409295 Chr.5: 40853501-40853501 on Build GRCh38
rs144328290 Chr.5: 40853462-40853462 on Build GRCh38
rs761948369 Chr.5: 40853582-40853582 on Build GRCh38
rs149994770 Chr.5: 40853596-40853596 on Build GRCh38
rs750806911 Chr.5: 40853594-40853594 on Build GRCh38
rs533019847 Chr.5: 40853434-40853434 on Build GRCh38
rs568392329 Chr.5: 40853526-40853526 on Build GRCh38
rs377069746 Chr.5: 40853580-40853580 on Build GRCh38
rs151228722 Chr.5: 40853503-40853503 on Build GRCh38
rs756884740 Chr.5: 40853626-40853626 on Build GRCh38
rs1009913002 Chr.5: 40853465-40853465 on Build GRCh38
rs746637092 Chr.5: 40853437-40853437 on Build GRCh38
rs763090706 Chr.5: 40853470-40853470 on Build GRCh38
rs139926562 Chr.5: 40853454-40853454 on Build GRCh38
rs780955753 Chr.5: 40853627-40853627 on Build GRCh38
rs754736417 Chr.5: 40853604-40853604 on Build GRCh38
rs747687670 Chr.5: 40853508-40853508 on Build GRCh38
rs923278939 Chr.5: 40853614-40853615 on Build GRCh38
rs1026238756 Chr.5: 40853528-40853528 on Build GRCh38
rs554211924 Chr.5: 40853566-40853566 on Build GRCh38
rs751779568 Chr.5: 40853481-40853481 on Build GRCh38
rs767838060 Chr.5: 40853488-40853488 on Build GRCh38
rs772699579 Chr.5: 40853431-40853431 on Build GRCh38
rs764112238 Chr.5: 40853561-40853561 on Build GRCh38
rs765211045 Chr.5: 40853456-40853456 on Build GRCh38
rs201397642 Chr.5: 40853595-40853595 on Build GRCh38
rs770610077 Chr.5: 40853545-40853545 on Build GRCh38
rs202113754 Chr.5: 40853538-40853538 on Build GRCh38
rs139253407 Chr.5: 40853514-40853514 on Build GRCh38
rs187034976 Chr.5: 40853424-40853424 on Build GRCh38
rs552212634 Chr.5: 40853448-40853448 on Build GRCh38

View Full Product Details