Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37310865-37311138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH4
Entrez Gene ID
27128
Gene Name
cytohesin 4
Gene Aliases
CYT4, DJ63G5.1, PSCD4
UniGene
Hs.170944
Gene Chromosome Location
Chr.22: 37282383-37315348 on Build GRCh38
Transcripts
NM_001318024.1 NM_013385.4 XM_011530147.2 XM_011530148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1327237 c.855C>G p.D285E Chr.22: 37311034-37311034 on Build GRCh38
444959 c.814C>T p.R272C Chr.22: 37310993-37310993 on Build GRCh38
3912750 c.862C>T p.L288F Chr.22: 37311041-37311041 on Build GRCh38
3150309 c.813C>G p.G271G Chr.22: 37310992-37310992 on Build GRCh38
3554140 c.843C>T p.F281F Chr.22: 37311022-37311022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370247166 Chr.22: 37311034-37311034 on Build GRCh38
rs56038904 Chr.22: 37310908-37310908 on Build GRCh38
rs770713425 Chr.22: 37311015-37311015 on Build GRCh38
rs374729035 Chr.22: 37311094-37311094 on Build GRCh38
rs1017389661 Chr.22: 37310993-37310993 on Build GRCh38
rs577150934 Chr.22: 37311032-37311032 on Build GRCh38
rs976127260 Chr.22: 37311014-37311014 on Build GRCh38
rs762501462 Chr.22: 37310952-37310952 on Build GRCh38
rs747476658 Chr.22: 37310947-37310949 on Build GRCh38
rs773606857 Chr.22: 37311044-37311044 on Build GRCh38
rs150393163 Chr.22: 37311052-37311052 on Build GRCh38
rs151256033 Chr.22: 37311004-37311004 on Build GRCh38
rs138472941 Chr.22: 37310983-37310983 on Build GRCh38
rs1030628409 Chr.22: 37310979-37310979 on Build GRCh38
rs750903557 Chr.22: 37310976-37310976 on Build GRCh38
rs774918484 Chr.22: 37310949-37310949 on Build GRCh38
rs757699186 Chr.22: 37310966-37310966 on Build GRCh38
rs752771550 Chr.22: 37310995-37310995 on Build GRCh38
rs781031003 Chr.22: 37311016-37311016 on Build GRCh38
rs766992258 Chr.22: 37311053-37311053 on Build GRCh38
rs949804879 Chr.22: 37311082-37311082 on Build GRCh38
rs777838924 Chr.22: 37311084-37311084 on Build GRCh38
rs56378525 Chr.22: 37311031-37311031 on Build GRCh38
rs55662193 Chr.22: 37311099-37311099 on Build GRCh38
rs758278339 Chr.22: 37311023-37311023 on Build GRCh38
rs779184298 Chr.22: 37310994-37310994 on Build GRCh38
rs779596631 Chr.22: 37311110-37311110 on Build GRCh38
rs913928663 Chr.22: 37311026-37311026 on Build GRCh38
rs754120263 Chr.22: 37310984-37310984 on Build GRCh38
rs11703243 Chr.22: 37310987-37310987 on Build GRCh38
rs548884809 Chr.22: 37310909-37310909 on Build GRCh38
rs767950459 Chr.22: 37310963-37310963 on Build GRCh38
rs201437695 Chr.22: 37311104-37311104 on Build GRCh38
rs188553120 Chr.22: 37311081-37311081 on Build GRCh38
rs768166234 Chr.22: 37311036-37311036 on Build GRCh38
rs751633533 Chr.22: 37311085-37311085 on Build GRCh38
rs1055334537 Chr.22: 37311076-37311076 on Build GRCh38
rs193101839 Chr.22: 37311097-37311097 on Build GRCh38
rs897503194 Chr.22: 37311092-37311092 on Build GRCh38
rs1043842647 Chr.22: 37310898-37310898 on Build GRCh38
rs752912934 Chr.22: 37311075-37311075 on Build GRCh38
rs751645635 Chr.22: 37311100-37311100 on Build GRCh38
rs376748528 Chr.22: 37311057-37311057 on Build GRCh38
rs759959838 Chr.22: 37311071-37311071 on Build GRCh38
rs1023448644 Chr.22: 37310989-37310989 on Build GRCh38
rs769431359 Chr.22: 37310942-37310942 on Build GRCh38
rs951921424 Chr.22: 37310917-37310917 on Build GRCh38
rs761143733 Chr.22: 37310981-37310981 on Build GRCh38
rs758461042 Chr.22: 37310996-37310996 on Build GRCh38

View Full Product Details