Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 157418342-157418597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTIP
Entrez Gene ID
9595
Gene Name
cytohesin 1 interacting protein
Gene Aliases
B3-1, CASP, CYBR, CYTHIP, HE, PSCDBP
UniGene
Hs.270
Gene Chromosome Location
Chr.2: 157412781-157444092 on Build GRCh38
Transcripts
NM_004288.4 XM_017005386.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3568178 c.601C>T p.R201C Chr.2: 157418535-157418535 on Build GRCh38
3568179 c.549A>T p.Q183H Chr.2: 157418587-157418587 on Build GRCh38
160315 c.594G>A p.Q198Q Chr.2: 157418542-157418542 on Build GRCh38
5993901 c.598C>G p.H200D Chr.2: 157418538-157418538 on Build GRCh38
3693871 c.592C>T p.Q198* Chr.2: 157418544-157418544 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs551932976 Chr.2: 157418494-157418494 on Build GRCh38
rs758057750 Chr.2: 157418563-157418563 on Build GRCh38
rs750278099 Chr.2: 157418450-157418450 on Build GRCh38
rs867301497 Chr.2: 157418526-157418526 on Build GRCh38
rs780965231 Chr.2: 157418433-157418433 on Build GRCh38
rs779498563 Chr.2: 157418567-157418567 on Build GRCh38
rs1002067623 Chr.2: 157418366-157418366 on Build GRCh38
rs777177600 Chr.2: 157418487-157418487 on Build GRCh38
rs778542165 Chr.2: 157418550-157418550 on Build GRCh38
rs563983594 Chr.2: 157418557-157418557 on Build GRCh38
rs199732380 Chr.2: 157418516-157418516 on Build GRCh38
rs749125160 Chr.2: 157418548-157418548 on Build GRCh38
rs183676401 Chr.2: 157418381-157418381 on Build GRCh38
rs770166816 Chr.2: 157418504-157418504 on Build GRCh38
rs769202933 Chr.2: 157418481-157418481 on Build GRCh38
rs138775935 Chr.2: 157418543-157418543 on Build GRCh38
rs1013642958 Chr.2: 157418400-157418400 on Build GRCh38
rs911833023 Chr.2: 157418438-157418438 on Build GRCh38
rs1026316404 Chr.2: 157418424-157418424 on Build GRCh38
rs754439491 Chr.2: 157418449-157418449 on Build GRCh38
rs760567533 Chr.2: 157418534-157418534 on Build GRCh38
rs766425231 Chr.2: 157418525-157418525 on Build GRCh38
rs950392830 Chr.2: 157418398-157418398 on Build GRCh38
rs144857541 Chr.2: 157418529-157418529 on Build GRCh38
rs763104365 Chr.2: 157418517-157418517 on Build GRCh38
rs763995680 Chr.2: 157418535-157418535 on Build GRCh38
rs747529148 Chr.2: 157418475-157418475 on Build GRCh38
rs749987868 Chr.2: 157418560-157418560 on Build GRCh38
rs530749560 Chr.2: 157418387-157418387 on Build GRCh38
rs753655443 Chr.2: 157418537-157418537 on Build GRCh38
rs780625465 Chr.2: 157418472-157418472 on Build GRCh38

View Full Product Details