Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100090361-100090592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPS6
Entrez Gene ID
10980
Gene Name
COP9 signalosome subunit 6
Gene Aliases
CSN6, MOV34-34KD
UniGene
Hs.15591
Gene Chromosome Location
Chr.7: 100088960-100092200 on Build GRCh38
Transcripts
NM_006833.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1093937 c.424-1G>A p.? Chr.7: 100090591-100090591 on Build GRCh38
3883925 c.401C>T p.S134L Chr.7: 100090465-100090465 on Build GRCh38
7337023 c.420G>T p.K140N Chr.7: 100090484-100090484 on Build GRCh38
7320537 c.364C>G p.L122V Chr.7: 100090428-100090428 on Build GRCh38
3951032 c.379A>G p.T127A Chr.7: 100090443-100090443 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756171637 Chr.7: 100090545-100090545 on Build GRCh38
rs754650627 Chr.7: 100090543-100090544 on Build GRCh38
rs145813985 Chr.7: 100090561-100090561 on Build GRCh38
rs773625233 Chr.7: 100090440-100090440 on Build GRCh38
rs749622502 Chr.7: 100090447-100090447 on Build GRCh38
rs1015845533 Chr.7: 100090391-100090391 on Build GRCh38
rs10277108 Chr.7: 100090521-100090521 on Build GRCh38
rs778320727 Chr.7: 100090551-100090551 on Build GRCh38
rs989560637 Chr.7: 100090548-100090548 on Build GRCh38
rs751770443 Chr.7: 100090516-100090516 on Build GRCh38
rs754431034 Chr.7: 100090542-100090542 on Build GRCh38
rs370286743 Chr.7: 100090465-100090465 on Build GRCh38
rs987701737 Chr.7: 100090499-100090499 on Build GRCh38
rs763548069 Chr.7: 100090563-100090563 on Build GRCh38
rs771471221 Chr.7: 100090449-100090449 on Build GRCh38
rs111371930 Chr.7: 100090398-100090398 on Build GRCh38
rs781277590 Chr.7: 100090526-100090526 on Build GRCh38
rs757011569 Chr.7: 100090503-100090503 on Build GRCh38
rs759319444 Chr.7: 100090460-100090460 on Build GRCh38
rs755049968 Chr.7: 100090395-100090395 on Build GRCh38
rs781393690 Chr.7: 100090412-100090412 on Build GRCh38
rs368635985 Chr.7: 100090556-100090556 on Build GRCh38
rs376570259 Chr.7: 100090452-100090452 on Build GRCh38
rs771713572 Chr.7: 100090558-100090558 on Build GRCh38
rs373454934 Chr.7: 100090566-100090567 on Build GRCh38
rs924081072 Chr.7: 100090510-100090510 on Build GRCh38
rs748193344 Chr.7: 100090430-100090430 on Build GRCh38
rs774141688 Chr.7: 100090451-100090451 on Build GRCh38
rs374263492 Chr.7: 100090532-100090532 on Build GRCh38
rs201795640 Chr.7: 100090392-100090392 on Build GRCh38
rs774797949 Chr.7: 100090557-100090557 on Build GRCh38
rs765086793 Chr.7: 100090504-100090504 on Build GRCh38
rs112342045 Chr.7: 100090397-100090397 on Build GRCh38
rs775024521 Chr.7: 100090560-100090560 on Build GRCh38
rs138487731 Chr.7: 100090475-100090475 on Build GRCh38
rs774997215 Chr.7: 100090466-100090466 on Build GRCh38
rs748220864 Chr.7: 100090536-100090536 on Build GRCh38
rs778231892 Chr.7: 100090548-100090551 on Build GRCh38
rs766190524 Chr.7: 100090476-100090476 on Build GRCh38
rs374891795 Chr.7: 100090433-100090433 on Build GRCh38
rs778538293 Chr.7: 100090552-100090552 on Build GRCh38

View Full Product Details