Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 871395-871656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD9
Entrez Gene ID
65980
Gene Name
bromodomain containing 9
Gene Aliases
LAVS3040, PRO9856
UniGene
Hs.449278
Gene Chromosome Location
Chr.5: 863735-892824 on Build GRCh38
Transcripts
NM_001009877.2 NM_001317951.1 NM_023924.4 NR_134296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618474 c.1108C>T p.P370S Chr.5: 871552-871552 on Build GRCh38
363389 c.1404G>T p.K468N Chr.5: 871544-871544 on Build GRCh38
4976473 c.1415A>T p.E472V Chr.5: 871533-871533 on Build GRCh38
363390 c.1116G>T p.K372N Chr.5: 871544-871544 on Build GRCh38
7186627 c.1391A>G p.N464S Chr.5: 871557-871557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753669540 Chr.5: 871568-871568 on Build GRCh38
rs765857516 Chr.5: 871567-871567 on Build GRCh38
rs142328423 Chr.5: 871432-871432 on Build GRCh38
rs749551621 Chr.5: 871501-871501 on Build GRCh38
rs780072998 Chr.5: 871477-871477 on Build GRCh38
rs773642121 Chr.5: 871494-871494 on Build GRCh38
rs897745800 Chr.5: 871582-871582 on Build GRCh38
rs1056904186 Chr.5: 871588-871588 on Build GRCh38
rs770650962 Chr.5: 871534-871534 on Build GRCh38
rs9313245 Chr.5: 871627-871627 on Build GRCh38
rs1022695807 Chr.5: 871620-871620 on Build GRCh38
rs376195552 Chr.5: 871602-871602 on Build GRCh38
rs369832383 Chr.5: 871586-871586 on Build GRCh38
rs376581330 Chr.5: 871553-871553 on Build GRCh38
rs760673235 Chr.5: 871566-871566 on Build GRCh38
rs372480384 Chr.5: 871618-871618 on Build GRCh38
rs373833562 Chr.5: 871614-871614 on Build GRCh38
rs770204062 Chr.5: 871493-871493 on Build GRCh38
rs748650818 Chr.5: 871484-871484 on Build GRCh38
rs375287367 Chr.5: 871628-871628 on Build GRCh38
rs1052677833 Chr.5: 871439-871439 on Build GRCh38
rs55744146 Chr.5: 871483-871483 on Build GRCh38
rs546159598 Chr.5: 871612-871612 on Build GRCh38
rs773829921 Chr.5: 871542-871542 on Build GRCh38
rs747142456 Chr.5: 871482-871482 on Build GRCh38
rs191778886 Chr.5: 871429-871429 on Build GRCh38
rs1013100240 Chr.5: 871619-871619 on Build GRCh38
rs1035008269 Chr.5: 871617-871617 on Build GRCh38
rs759171078 Chr.5: 871549-871549 on Build GRCh38
rs775632564 Chr.5: 871557-871557 on Build GRCh38

View Full Product Details