Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 36926114-36926339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2RB
Entrez Gene ID
1439
Gene Name
colony stimulating factor 2 receptor beta common subunit
Gene Aliases
CD131, CDw131, IL3RB, IL5RB, SMDP5
UniGene
Hs.592192
Gene Chromosome Location
Chr.22: 36913574-36940449 on Build GRCh38
Transcripts
NM_000395.2 XM_005261340.3 XM_011529903.2 XM_011529904.2 XM_011529905.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
295130 c.364C>T p.R122W Chr.22: 36926150-36926150 on Build GRCh38
356126 c.333C>G p.F111L Chr.22: 36926119-36926119 on Build GRCh38
5393551 c.346G>A p.D116N Chr.22: 36926132-36926132 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs544454928 Chr.22: 36926286-36926286 on Build GRCh38
rs775669271 Chr.22: 36926146-36926146 on Build GRCh38
rs963204212 Chr.22: 36926306-36926306 on Build GRCh38
rs768923587 Chr.22: 36926213-36926213 on Build GRCh38
rs754500967 Chr.22: 36926175-36926175 on Build GRCh38
rs959748683 Chr.22: 36926247-36926247 on Build GRCh38
rs780432541 Chr.22: 36926206-36926206 on Build GRCh38
rs375819512 Chr.22: 36926151-36926151 on Build GRCh38
rs749366022 Chr.22: 36926212-36926212 on Build GRCh38
rs757533889 Chr.22: 36926197-36926197 on Build GRCh38
rs141354922 Chr.22: 36926158-36926158 on Build GRCh38
rs752179178 Chr.22: 36926191-36926191 on Build GRCh38
rs907687376 Chr.22: 36926292-36926292 on Build GRCh38
rs755285531 Chr.22: 36926173-36926173 on Build GRCh38
rs761508909 Chr.22: 36926153-36926153 on Build GRCh38
rs373203342 Chr.22: 36926204-36926204 on Build GRCh38
rs781269565 Chr.22: 36926137-36926137 on Build GRCh38
rs781569278 Chr.22: 36926200-36926200 on Build GRCh38
rs766052297 Chr.22: 36926169-36926169 on Build GRCh38
rs748164035 Chr.22: 36926227-36926227 on Build GRCh38
rs146302338 Chr.22: 36926159-36926159 on Build GRCh38
rs369381768 Chr.22: 36926188-36926188 on Build GRCh38
rs986050603 Chr.22: 36926268-36926268 on Build GRCh38
rs746247362 Chr.22: 36926201-36926201 on Build GRCh38
rs770160808 Chr.22: 36926143-36926143 on Build GRCh38
rs746144466 Chr.22: 36926139-36926139 on Build GRCh38
rs202034054 Chr.22: 36926150-36926150 on Build GRCh38

View Full Product Details