Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171278672-171278927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO1
Entrez Gene ID
2326
Gene Name
flavin containing monooxygenase 1
Gene Aliases
-
UniGene
Hs.1424
Gene Chromosome Location
Chr.1: 171248471-171285978 on Build GRCh38
Transcripts
NM_001282692.1 NM_001282693.1 NM_001282694.1 NM_002021.2 XM_005245037.3 XM_005245038.3 XM_006711241.3 XM_006711242.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1252624 c.552C>T p.D184D Chr.1: 171278796-171278796 on Build GRCh38
6541234 c.619G>T p.A207S Chr.1: 171278863-171278863 on Build GRCh38
3478279 c.562C>T p.L188F Chr.1: 171278806-171278806 on Build GRCh38
1689168 c.578G>A p.G193E Chr.1: 171278822-171278822 on Build GRCh38
5551789 c.592G>A p.D198N Chr.1: 171278836-171278836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767949068 Chr.1: 171278877-171278877 on Build GRCh38
rs770843838 Chr.1: 171278737-171278737 on Build GRCh38
rs751946528 Chr.1: 171278762-171278762 on Build GRCh38
rs752154064 Chr.1: 171278785-171278785 on Build GRCh38
rs770751927 Chr.1: 171278814-171278814 on Build GRCh38
rs967201028 Chr.1: 171278787-171278787 on Build GRCh38
rs146934200 Chr.1: 171278864-171278864 on Build GRCh38
rs780996075 Chr.1: 171278836-171278836 on Build GRCh38
rs751131056 Chr.1: 171278901-171278901 on Build GRCh38
rs200537259 Chr.1: 171278747-171278747 on Build GRCh38
rs778509179 Chr.1: 171278784-171278784 on Build GRCh38
rs745756826 Chr.1: 171278839-171278839 on Build GRCh38
rs746848448 Chr.1: 171278734-171278734 on Build GRCh38
rs145849541 Chr.1: 171278760-171278760 on Build GRCh38
rs1002816863 Chr.1: 171278771-171278771 on Build GRCh38
rs775568390 Chr.1: 171278860-171278860 on Build GRCh38
rs28360407 Chr.1: 171278719-171278719 on Build GRCh38
rs746796373 Chr.1: 171278809-171278809 on Build GRCh38
rs778601183 Chr.1: 171278710-171278710 on Build GRCh38
rs371512929 Chr.1: 171278879-171278879 on Build GRCh38
rs767380447 Chr.1: 171278826-171278826 on Build GRCh38
rs769824265 Chr.1: 171278751-171278751 on Build GRCh38
rs377169079 Chr.1: 171278765-171278765 on Build GRCh38
rs369942179 Chr.1: 171278754-171278754 on Build GRCh38
rs763770203 Chr.1: 171278900-171278900 on Build GRCh38
rs756714478 Chr.1: 171278782-171278782 on Build GRCh38
rs201784982 Chr.1: 171278738-171278738 on Build GRCh38
rs147697313 Chr.1: 171278828-171278828 on Build GRCh38
rs766862433 Chr.1: 171278880-171278880 on Build GRCh38
rs747791324 Chr.1: 171278716-171278716 on Build GRCh38
rs200847805 Chr.1: 171278717-171278717 on Build GRCh38
rs777350045 Chr.1: 171278802-171278802 on Build GRCh38
rs757896074 Chr.1: 171278895-171278895 on Build GRCh38
rs761015991 Chr.1: 171278880-171278881 on Build GRCh38
rs752403491 Chr.1: 171278833-171278836 on Build GRCh38
rs866118693 Chr.1: 171278816-171278816 on Build GRCh38
rs367730433 Chr.1: 171278865-171278865 on Build GRCh38
rs977693406 Chr.1: 171278806-171278806 on Build GRCh38
rs752240581 Chr.1: 171278882-171278882 on Build GRCh38
rs372785457 Chr.1: 171278764-171278764 on Build GRCh38
rs769733946 Chr.1: 171278842-171278842 on Build GRCh38
rs754577914 Chr.1: 171278702-171278702 on Build GRCh38
rs535797940 Chr.1: 171278862-171278862 on Build GRCh38
rs764218641 Chr.1: 171278882-171278883 on Build GRCh38

View Full Product Details