Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 59339871-59340132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGGY
Entrez Gene ID
55277
Gene Name
FGGY carbohydrate kinase domain containing
Gene Aliases
-
UniGene
Hs.444301
Gene Chromosome Location
Chr.1: 59296405-59762730 on Build GRCh38
Transcripts
NM_001113411.1 NM_001244714.1 NM_018291.3 NR_103473.1 XM_011541730.1 XM_011541731.1 XM_011541732.1 XM_011541733.1 XM_011541735.1 XM_011541736.2 XM_017001643.1 XM_017001644.1 XM_017001645.1 XM_017001646.1 XM_017001647.1 XM_017001648.1 XM_017001649.1 XM_017001650.1 XM_017001651.1 XM_017001652.1 XM_017001653.1 XM_017001654.1 XM_017001655.1 XM_017001656.1 XM_017001657.1 XM_017001658.1 XM_017001659.1 XM_017001660.1 XM_017001661.1 XM_017001662.1 XM_017001663.1 XM_017001664.1 XM_017001665.1 XM_017001666.1 XM_017001667.1 XM_017001668.1 XM_017001669.1 XM_017001670.1 XM_017001671.1 XM_017001672.1 XM_017001673.1 XM_017001674.1 XM_017001675.1 XM_017001676.1 XM_017001679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4535065 c.214G>A p.G72R Chr.1: 59339970-59339970 on Build GRCh38
5381323 c.254C>T p.A85V Chr.1: 59340010-59340010 on Build GRCh38
3491359 c.236G>A p.R79Q Chr.1: 59339992-59339992 on Build GRCh38
6460584 c.255C>T p.A85A Chr.1: 59340011-59340011 on Build GRCh38
3491358 c.235C>T p.R79* Chr.1: 59339991-59339991 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775881455 Chr.1: 59339914-59339914 on Build GRCh38
rs769544537 Chr.1: 59339956-59339957 on Build GRCh38
rs945771884 Chr.1: 59340098-59340098 on Build GRCh38
rs762132178 Chr.1: 59340053-59340053 on Build GRCh38
rs973666753 Chr.1: 59339921-59339921 on Build GRCh38
rs370557992 Chr.1: 59339943-59339943 on Build GRCh38
rs374916903 Chr.1: 59340013-59340013 on Build GRCh38
rs764668040 Chr.1: 59340105-59340105 on Build GRCh38
rs759164028 Chr.1: 59340072-59340072 on Build GRCh38
rs866400634 Chr.1: 59339958-59339958 on Build GRCh38
rs1027698261 Chr.1: 59339988-59339988 on Build GRCh38
rs747560457 Chr.1: 59340101-59340102 on Build GRCh38
rs746050054 Chr.1: 59340034-59340034 on Build GRCh38
rs993404038 Chr.1: 59340015-59340015 on Build GRCh38
rs748040209 Chr.1: 59339991-59339991 on Build GRCh38
rs762811097 Chr.1: 59339970-59339970 on Build GRCh38
rs773047064 Chr.1: 59339967-59339967 on Build GRCh38
rs370879908 Chr.1: 59340048-59340048 on Build GRCh38
rs184449304 Chr.1: 59340041-59340041 on Build GRCh38
rs753384391 Chr.1: 59339955-59339955 on Build GRCh38
rs935974817 Chr.1: 59340103-59340103 on Build GRCh38
rs776974816 Chr.1: 59340030-59340030 on Build GRCh38
rs766009504 Chr.1: 59339951-59339951 on Build GRCh38
rs983338644 Chr.1: 59340002-59340002 on Build GRCh38
rs774763998 Chr.1: 59340049-59340049 on Build GRCh38
rs868444510 Chr.1: 59340045-59340045 on Build GRCh38
rs755822091 Chr.1: 59339922-59339922 on Build GRCh38
rs181105009 Chr.1: 59339966-59339966 on Build GRCh38
rs751862549 Chr.1: 59339920-59339920 on Build GRCh38
rs913013470 Chr.1: 59340078-59340078 on Build GRCh38
rs746800060 Chr.1: 59340011-59340011 on Build GRCh38
rs929484508 Chr.1: 59340014-59340014 on Build GRCh38
rs551303292 Chr.1: 59339965-59339965 on Build GRCh38
rs778447469 Chr.1: 59339982-59339982 on Build GRCh38
rs752242420 Chr.1: 59340109-59340109 on Build GRCh38
rs765819706 Chr.1: 59339908-59339908 on Build GRCh38
rs763380259 Chr.1: 59340047-59340047 on Build GRCh38
rs777772308 Chr.1: 59340000-59340000 on Build GRCh38
rs775568452 Chr.1: 59340046-59340046 on Build GRCh38
rs566794456 Chr.1: 59339917-59339917 on Build GRCh38
rs758298995 Chr.1: 59339992-59339992 on Build GRCh38
rs753466414 Chr.1: 59340064-59340064 on Build GRCh38
rs754470884 Chr.1: 59339971-59339971 on Build GRCh38
rs764443621 Chr.1: 59339916-59339916 on Build GRCh38

View Full Product Details