Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 230839745-230840006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf198
Entrez Gene ID
84886
Gene Name
chromosome 1 open reading frame 198
Gene Aliases
-
UniGene
Hs.520494
Gene Chromosome Location
Chr.1: 230837119-230869589 on Build GRCh38
Transcripts
NM_001136494.1 NM_001136495.1 NM_032800.2 XM_017002599.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6541950 c.935C>A p.S312* Chr.1: 230839901-230839901 on Build GRCh38
5454109 c.931A>G p.S311G Chr.1: 230839905-230839905 on Build GRCh38
4667361 c.945C>T p.V315V Chr.1: 230839891-230839891 on Build GRCh38
4029396 c.946G>A p.V316I Chr.1: 230839890-230839890 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs946418095 Chr.1: 230839895-230839895 on Build GRCh38
rs747733084 Chr.1: 230839948-230839948 on Build GRCh38
rs774576054 Chr.1: 230839832-230839832 on Build GRCh38
rs192101698 Chr.1: 230839817-230839817 on Build GRCh38
rs187402722 Chr.1: 230839802-230839802 on Build GRCh38
rs941495090 Chr.1: 230839972-230839972 on Build GRCh38
rs772297312 Chr.1: 230839940-230839940 on Build GRCh38
rs755868627 Chr.1: 230839924-230839924 on Build GRCh38
rs764514710 Chr.1: 230839879-230839879 on Build GRCh38
rs777159565 Chr.1: 230839880-230839880 on Build GRCh38
rs766038551 Chr.1: 230839891-230839891 on Build GRCh38
rs777704326 Chr.1: 230839815-230839815 on Build GRCh38
rs769525848 Chr.1: 230839955-230839956 on Build GRCh38
rs746136611 Chr.1: 230839834-230839834 on Build GRCh38
rs955979220 Chr.1: 230839778-230839778 on Build GRCh38
rs757656014 Chr.1: 230839918-230839918 on Build GRCh38
rs745935017 Chr.1: 230839938-230839938 on Build GRCh38
rs372998173 Chr.1: 230839922-230839922 on Build GRCh38
rs996593701 Chr.1: 230839946-230839946 on Build GRCh38
rs542332737 Chr.1: 230839888-230839890 on Build GRCh38
rs944510577 Chr.1: 230839790-230839790 on Build GRCh38
rs529493243 Chr.1: 230839890-230839890 on Build GRCh38
rs111322053 Chr.1: 230839947-230839948 on Build GRCh38
rs534935093 Chr.1: 230839974-230839974 on Build GRCh38
rs375958334 Chr.1: 230839831-230839831 on Build GRCh38
rs746432533 Chr.1: 230839961-230839961 on Build GRCh38
rs369345382 Chr.1: 230839849-230839849 on Build GRCh38
rs779004058 Chr.1: 230839935-230839935 on Build GRCh38
rs761042620 Chr.1: 230839860-230839860 on Build GRCh38
rs746964070 Chr.1: 230839836-230839848 on Build GRCh38
rs183137802 Chr.1: 230839963-230839963 on Build GRCh38
rs11538432 Chr.1: 230839847-230839847 on Build GRCh38
rs904441244 Chr.1: 230839965-230839965 on Build GRCh38
rs777601706 Chr.1: 230839929-230839929 on Build GRCh38
rs1048192496 Chr.1: 230839919-230839919 on Build GRCh38
rs371003599 Chr.1: 230839941-230839941 on Build GRCh38
rs775892753 Chr.1: 230839858-230839858 on Build GRCh38
rs1043746090 Chr.1: 230839911-230839911 on Build GRCh38
rs992621479 Chr.1: 230839785-230839785 on Build GRCh38
rs377377090 Chr.1: 230839931-230839931 on Build GRCh38
rs754741512 Chr.1: 230839917-230839917 on Build GRCh38
rs920815935 Chr.1: 230839824-230839824 on Build GRCh38
rs757163285 Chr.1: 230839934-230839934 on Build GRCh38
rs751086607 Chr.1: 230839907-230839907 on Build GRCh38

View Full Product Details