Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 59837125-59837398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIF
Entrez Gene ID
2694
Gene Name
gastric intrinsic factor
Gene Aliases
IF, IFMH, INF, TCN3
UniGene
Hs.110014
Gene Chromosome Location
Chr.11: 59829268-59845529 on Build GRCh38
Transcripts
NM_005142.2 XM_011544939.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6133431 c.848T>G p.V283G Chr.11: 59837197-59837197 on Build GRCh38
2035751 c.836C>A p.T279K Chr.11: 59837209-59837209 on Build GRCh38
2035753 c.794C>T p.P265L Chr.11: 59837251-59837251 on Build GRCh38
6248189 c.831C>A p.G277G Chr.11: 59837214-59837214 on Build GRCh38
3809695 c.723A>C p.K241N Chr.11: 59837322-59837322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35867471 Chr.11: 59837281-59837281 on Build GRCh38
rs533802342 Chr.11: 59837198-59837198 on Build GRCh38
rs149355084 Chr.11: 59837342-59837342 on Build GRCh38
rs773563039 Chr.11: 59837326-59837326 on Build GRCh38
rs773699225 Chr.11: 59837150-59837150 on Build GRCh38
rs769027647 Chr.11: 59837160-59837160 on Build GRCh38
rs748783434 Chr.11: 59837248-59837248 on Build GRCh38
rs377432418 Chr.11: 59837295-59837295 on Build GRCh38
rs138504371 Chr.11: 59837216-59837216 on Build GRCh38
rs781140362 Chr.11: 59837256-59837256 on Build GRCh38
rs774885583 Chr.11: 59837302-59837302 on Build GRCh38
rs755030986 Chr.11: 59837250-59837250 on Build GRCh38
rs979617516 Chr.11: 59837258-59837258 on Build GRCh38
rs756139879 Chr.11: 59837267-59837267 on Build GRCh38
rs139320672 Chr.11: 59837294-59837294 on Build GRCh38
rs777684435 Chr.11: 59837275-59837275 on Build GRCh38
rs948418280 Chr.11: 59837349-59837349 on Build GRCh38
rs766656335 Chr.11: 59837220-59837220 on Build GRCh38
rs767581371 Chr.11: 59837366-59837366 on Build GRCh38
rs777137202 Chr.11: 59837186-59837186 on Build GRCh38
rs542431882 Chr.11: 59837296-59837296 on Build GRCh38
rs886048402 Chr.11: 59837191-59837191 on Build GRCh38
rs983078302 Chr.11: 59837251-59837251 on Build GRCh38
rs752643448 Chr.11: 59837257-59837257 on Build GRCh38
rs371638397 Chr.11: 59837190-59837190 on Build GRCh38
rs1048065881 Chr.11: 59837336-59837336 on Build GRCh38
rs886048403 Chr.11: 59837211-59837211 on Build GRCh38
rs751581445 Chr.11: 59837247-59837247 on Build GRCh38
rs142495062 Chr.11: 59837268-59837268 on Build GRCh38
rs770042905 Chr.11: 59837344-59837344 on Build GRCh38
rs759151652 Chr.11: 59837157-59837157 on Build GRCh38
rs201261367 Chr.11: 59837310-59837310 on Build GRCh38
rs762285472 Chr.11: 59837304-59837304 on Build GRCh38
rs759712731 Chr.11: 59837348-59837348 on Build GRCh38

View Full Product Details