Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18246608-18246855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLII
Entrez Gene ID
2314
Gene Name
FLII, actin remodeling protein
Gene Aliases
FLI, FLIL, Fli1
UniGene
Hs.513984
Gene Chromosome Location
Chr.17: 18244815-18258916 on Build GRCh38
Transcripts
NM_001256264.1 NM_001256265.1 NM_002018.3 XM_005256555.3 XM_005256556.3 XM_005256558.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6664645 c.2823G>T p.W941C Chr.17: 18246822-18246822 on Build GRCh38
976262 c.2853G>T p.K951N Chr.17: 18246792-18246792 on Build GRCh38
6664646 c.2949C>T p.I983I Chr.17: 18246696-18246696 on Build GRCh38
3889392 c.3003C>T p.T1001T Chr.17: 18246642-18246642 on Build GRCh38
3988820 c.3024C>T p.F1008F Chr.17: 18246621-18246621 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539543836 Chr.17: 18246713-18246713 on Build GRCh38
rs935917289 Chr.17: 18246780-18246780 on Build GRCh38
rs750070579 Chr.17: 18246806-18246806 on Build GRCh38
rs774531113 Chr.17: 18246768-18246768 on Build GRCh38
rs765490816 Chr.17: 18246683-18246683 on Build GRCh38
rs202103078 Chr.17: 18246804-18246804 on Build GRCh38
rs74508005 Chr.17: 18246632-18246632 on Build GRCh38
rs755944555 Chr.17: 18246780-18246782 on Build GRCh38
rs752686813 Chr.17: 18246777-18246777 on Build GRCh38
rs765214163 Chr.17: 18246725-18246725 on Build GRCh38
rs754839621 Chr.17: 18246711-18246725 on Build GRCh38
rs908017544 Chr.17: 18246639-18246639 on Build GRCh38
rs924522553 Chr.17: 18246810-18246810 on Build GRCh38
rs747523576 Chr.17: 18246649-18246649 on Build GRCh38
rs141780549 Chr.17: 18246834-18246834 on Build GRCh38
rs752614270 Chr.17: 18246794-18246794 on Build GRCh38
rs745323823 Chr.17: 18246722-18246722 on Build GRCh38
rs751594817 Chr.17: 18246686-18246686 on Build GRCh38
rs778760276 Chr.17: 18246803-18246803 on Build GRCh38
rs778516769 Chr.17: 18246721-18246721 on Build GRCh38
rs766497862 Chr.17: 18246685-18246685 on Build GRCh38
rs754927239 Chr.17: 18246693-18246693 on Build GRCh38
rs546354452 Chr.17: 18246748-18246748 on Build GRCh38
rs781090764 Chr.17: 18246726-18246728 on Build GRCh38
rs777753109 Chr.17: 18246796-18246798 on Build GRCh38
rs762318935 Chr.17: 18246666-18246666 on Build GRCh38
rs767578353 Chr.17: 18246774-18246774 on Build GRCh38
rs781775406 Chr.17: 18246735-18246735 on Build GRCh38
rs559248106 Chr.17: 18246718-18246718 on Build GRCh38
rs756957111 Chr.17: 18246798-18246798 on Build GRCh38
rs201095894 Chr.17: 18246751-18246751 on Build GRCh38
rs1026565854 Chr.17: 18246753-18246753 on Build GRCh38
rs753542412 Chr.17: 18246714-18246714 on Build GRCh38
rs767490128 Chr.17: 18246695-18246695 on Build GRCh38
rs200248186 Chr.17: 18246766-18246766 on Build GRCh38
rs779700268 Chr.17: 18246816-18246816 on Build GRCh38
rs748682570 Chr.17: 18246818-18246818 on Build GRCh38
rs200455487 Chr.17: 18246772-18246772 on Build GRCh38
rs769246541 Chr.17: 18246662-18246662 on Build GRCh38
rs752596764 Chr.17: 18246698-18246698 on Build GRCh38
rs117072920 Chr.17: 18246664-18246664 on Build GRCh38
rs555286267 Chr.17: 18246797-18246797 on Build GRCh38
rs756002975 Chr.17: 18246711-18246711 on Build GRCh38
rs139095003 Chr.17: 18246774-18246776 on Build GRCh38
rs201937124 Chr.17: 18246740-18246740 on Build GRCh38
rs200030922 Chr.17: 18246729-18246729 on Build GRCh38
rs778218444 Chr.17: 18246836-18246836 on Build GRCh38
rs150392959 Chr.17: 18246648-18246648 on Build GRCh38
rs773394547 Chr.17: 18246762-18246762 on Build GRCh38
rs932068935 Chr.17: 18246755-18246755 on Build GRCh38
rs763077210 Chr.17: 18246684-18246684 on Build GRCh38
rs566163422 Chr.17: 18246764-18246764 on Build GRCh38
rs575181558 Chr.17: 18246813-18246813 on Build GRCh38
rs751804874 Chr.17: 18246709-18246711 on Build GRCh38
rs760471288 Chr.17: 18246782-18246782 on Build GRCh38

View Full Product Details