Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 74947993-74948252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf40
Entrez Gene ID
55071
Gene Name
chromosome 9 open reading frame 40
Gene Aliases
-
UniGene
Hs.532296
Gene Chromosome Location
Chr.9: 74946583-74952886 on Build GRCh38
Transcripts
NM_017998.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1462966 c.470G>T p.R157M Chr.9: 74948163-74948163 on Build GRCh38
150620 c.462G>A p.Q154Q Chr.9: 74948171-74948171 on Build GRCh38
3658244 c.544C>T p.Q182* Chr.9: 74948089-74948089 on Build GRCh38
6056710 c.487A>G p.I163V Chr.9: 74948146-74948146 on Build GRCh38
4668165 c.555T>C p.N185N Chr.9: 74948078-74948078 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982600574 Chr.9: 74948170-74948170 on Build GRCh38
rs765247019 Chr.9: 74948029-74948029 on Build GRCh38
rs2769058 Chr.9: 74948171-74948171 on Build GRCh38
rs747294638 Chr.9: 74948041-74948041 on Build GRCh38
rs763770773 Chr.9: 74948122-74948122 on Build GRCh38
rs753126147 Chr.9: 74948086-74948086 on Build GRCh38
rs754778387 Chr.9: 74948181-74948181 on Build GRCh38
rs748288518 Chr.9: 74948047-74948047 on Build GRCh38
rs186036248 Chr.9: 74948158-74948158 on Build GRCh38
rs755265761 Chr.9: 74948045-74948045 on Build GRCh38
rs752840864 Chr.9: 74948202-74948202 on Build GRCh38
rs147106981 Chr.9: 74948052-74948052 on Build GRCh38
rs951294139 Chr.9: 74948167-74948167 on Build GRCh38
rs773817891 Chr.9: 74948082-74948082 on Build GRCh38
rs377278085 Chr.9: 74948210-74948210 on Build GRCh38
rs201273146 Chr.9: 74948108-74948108 on Build GRCh38
rs781585355 Chr.9: 74948189-74948189 on Build GRCh38
rs756684380 Chr.9: 74948028-74948028 on Build GRCh38
rs1034444854 Chr.9: 74948143-74948143 on Build GRCh38
rs369913991 Chr.9: 74948036-74948036 on Build GRCh38
rs767956703 Chr.9: 74948031-74948031 on Build GRCh38
rs886075914 Chr.9: 74948033-74948033 on Build GRCh38
rs374482931 Chr.9: 74948109-74948109 on Build GRCh38
rs549664253 Chr.9: 74948131-74948131 on Build GRCh38
rs764625310 Chr.9: 74948151-74948151 on Build GRCh38
rs374853709 Chr.9: 74948037-74948037 on Build GRCh38
rs767300906 Chr.9: 74948214-74948215 on Build GRCh38
rs761439243 Chr.9: 74948132-74948132 on Build GRCh38
rs759834017 Chr.9: 74948112-74948112 on Build GRCh38
rs781497162 Chr.9: 74948046-74948046 on Build GRCh38
rs753353619 Chr.9: 74948022-74948022 on Build GRCh38
rs377615362 Chr.9: 74948153-74948153 on Build GRCh38
rs369640750 Chr.9: 74948166-74948166 on Build GRCh38
rs1024795692 Chr.9: 74948180-74948180 on Build GRCh38
rs749723607 Chr.9: 74948106-74948106 on Build GRCh38

View Full Product Details