Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39642738-39642981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1I
Entrez Gene ID
8911
Gene Name
calcium voltage-gated channel subunit alpha1 I
Gene Aliases
Cav3.3, ca(v)3.3
UniGene
Hs.125116
Gene Chromosome Location
Chr.22: 39570753-39689737 on Build GRCh38
Transcripts
NM_001003406.1 NM_021096.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1721165 c.1137C>T p.I379I Chr.22: 39642877-39642877 on Build GRCh38
726260 c.1118A>G p.Y373C Chr.22: 39642858-39642858 on Build GRCh38
6249854 c.1089G>T p.M363I Chr.22: 39642829-39642829 on Build GRCh38
3554611 c.1072G>A p.G358S Chr.22: 39642812-39642812 on Build GRCh38
1196012 c.1089G>A p.M363I Chr.22: 39642829-39642829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751904962 Chr.22: 39642933-39642933 on Build GRCh38
rs200241888 Chr.22: 39642788-39642788 on Build GRCh38
rs201234915 Chr.22: 39642771-39642771 on Build GRCh38
rs60820415 Chr.22: 39642790-39642790 on Build GRCh38
rs996254696 Chr.22: 39642805-39642805 on Build GRCh38
rs768526757 Chr.22: 39642851-39642851 on Build GRCh38
rs776584119 Chr.22: 39642862-39642862 on Build GRCh38
rs975500775 Chr.22: 39642775-39642775 on Build GRCh38
rs758764945 Chr.22: 39642756-39642756 on Build GRCh38
rs57495596 Chr.22: 39642835-39642835 on Build GRCh38
rs56689202 Chr.22: 39642906-39642906 on Build GRCh38
rs1054261467 Chr.22: 39642898-39642898 on Build GRCh38
rs372816192 Chr.22: 39642787-39642787 on Build GRCh38
rs766578987 Chr.22: 39642929-39642929 on Build GRCh38
rs753879577 Chr.22: 39642770-39642770 on Build GRCh38
rs957395112 Chr.22: 39642837-39642837 on Build GRCh38
rs746788319 Chr.22: 39642850-39642850 on Build GRCh38
rs886708235 Chr.22: 39642777-39642777 on Build GRCh38
rs546117537 Chr.22: 39642925-39642925 on Build GRCh38
rs935870675 Chr.22: 39642877-39642877 on Build GRCh38
rs773436321 Chr.22: 39642924-39642924 on Build GRCh38
rs781699909 Chr.22: 39642782-39642782 on Build GRCh38
rs765233536 Chr.22: 39642908-39642908 on Build GRCh38
rs771841014 Chr.22: 39642827-39642827 on Build GRCh38
rs755345849 Chr.22: 39642937-39642937 on Build GRCh38
rs1020173057 Chr.22: 39642911-39642911 on Build GRCh38
rs61506789 Chr.22: 39642774-39642774 on Build GRCh38
rs750751457 Chr.22: 39642789-39642789 on Build GRCh38

View Full Product Details