Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96982351-96982576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf12
Entrez Gene ID
26148
Gene Name
chromosome 10 open reading frame 12
Gene Aliases
-
UniGene
Hs.427927
Gene Chromosome Location
Chr.10: 96981284-96985828 on Build GRCh38
Transcripts
NM_015652.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7424896 c.1097G>C p.S366T Chr.10: 96982487-96982487 on Build GRCh38
4568318 c.1083T>G p.L361L Chr.10: 96982473-96982473 on Build GRCh38
6890998 c.1089C>T p.S363S Chr.10: 96982479-96982479 on Build GRCh38
921952 c.1068G>T p.Q356H Chr.10: 96982458-96982458 on Build GRCh38
6613793 c.1036C>T p.L346L Chr.10: 96982426-96982426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762555085 Chr.10: 96982542-96982542 on Build GRCh38
rs144441056 Chr.10: 96982494-96982494 on Build GRCh38
rs775587920 Chr.10: 96982421-96982421 on Build GRCh38
rs199784223 Chr.10: 96982506-96982506 on Build GRCh38
rs753515408 Chr.10: 96982440-96982440 on Build GRCh38
rs183640777 Chr.10: 96982436-96982436 on Build GRCh38
rs750100651 Chr.10: 96982537-96982537 on Build GRCh38
rs972399465 Chr.10: 96982415-96982415 on Build GRCh38
rs748721986 Chr.10: 96982467-96982467 on Build GRCh38
rs763951095 Chr.10: 96982437-96982437 on Build GRCh38
rs750049126 Chr.10: 96982448-96982448 on Build GRCh38
rs762316678 Chr.10: 96982493-96982493 on Build GRCh38
rs542265504 Chr.10: 96982391-96982391 on Build GRCh38
rs774710171 Chr.10: 96982409-96982409 on Build GRCh38
rs770219418 Chr.10: 96982372-96982372 on Build GRCh38
rs920115011 Chr.10: 96982432-96982432 on Build GRCh38
rs746036515 Chr.10: 96982499-96982499 on Build GRCh38
rs1046422181 Chr.10: 96982488-96982488 on Build GRCh38
rs761556275 Chr.10: 96982532-96982532 on Build GRCh38
rs770116436 Chr.10: 96982441-96982441 on Build GRCh38
rs150528336 Chr.10: 96982444-96982444 on Build GRCh38
rs776630981 Chr.10: 96982526-96982526 on Build GRCh38
rs149255891 Chr.10: 96982468-96982468 on Build GRCh38
rs763358038 Chr.10: 96982536-96982536 on Build GRCh38
rs1012167730 Chr.10: 96982373-96982373 on Build GRCh38
rs139771197 Chr.10: 96982495-96982495 on Build GRCh38
rs778214101 Chr.10: 96982475-96982475 on Build GRCh38
rs373519273 Chr.10: 96982510-96982510 on Build GRCh38
rs115140616 Chr.10: 96982461-96982461 on Build GRCh38
rs759729191 Chr.10: 96982413-96982413 on Build GRCh38
rs765880547 Chr.10: 96982544-96982544 on Build GRCh38
rs201494751 Chr.10: 96982417-96982417 on Build GRCh38
rs878915612 Chr.10: 96982397-96982399 on Build GRCh38
rs568711516 Chr.10: 96982385-96982385 on Build GRCh38
rs750977287 Chr.10: 96982545-96982545 on Build GRCh38
rs142473396 Chr.10: 96982400-96982400 on Build GRCh38

View Full Product Details