Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 35186747-35186996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD44
Entrez Gene ID
960
Gene Name
CD44 molecule (Indian blood group)
Gene Aliases
CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1
UniGene
Hs.502328
Gene Chromosome Location
Chr.11: 35138870-35232402 on Build GRCh38
Transcripts
NM_000610.3 NM_001001389.1 NM_001001390.1 NM_001001391.1 NM_001001392.1 NM_001202555.1 NM_001202556.1 NM_001202557.1 XM_005253231.2 XM_005253232.2 XM_005253233.2 XM_005253234.2 XM_005253235.2 XM_005253238.2 XM_005253239.2 XM_005253240.2 XM_006718388.1 XM_006718390.3 XM_011520482.1 XM_011520483.1 XM_011520484.1 XM_011520485.1 XM_011520486.1 XM_011520487.2 XM_011520488.1 XM_011520489.2 XM_017018583.1 XM_017018584.1 XM_017018585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466776 c.436+2T>A p.? Chr.11: 35186902-35186902 on Build GRCh38
5475501 c.415T>G p.F139V Chr.11: 35186879-35186879 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748532837 Chr.11: 35186919-35186919 on Build GRCh38
rs766188974 Chr.11: 35186819-35186819 on Build GRCh38
rs762633485 Chr.11: 35186811-35186811 on Build GRCh38
rs751897249 Chr.11: 35186913-35186913 on Build GRCh38
rs374064563 Chr.11: 35186956-35186956 on Build GRCh38
rs759446399 Chr.11: 35186828-35186828 on Build GRCh38
rs762951199 Chr.11: 35186910-35186910 on Build GRCh38
rs757360424 Chr.11: 35186813-35186813 on Build GRCh38
rs780293069 Chr.11: 35186911-35186911 on Build GRCh38
rs111990873 Chr.11: 35186820-35186820 on Build GRCh38
rs770058920 Chr.11: 35186812-35186813 on Build GRCh38
rs748876826 Chr.11: 35186947-35186947 on Build GRCh38
rs765163014 Chr.11: 35186877-35186877 on Build GRCh38
rs903733245 Chr.11: 35186810-35186810 on Build GRCh38
rs200524971 Chr.11: 35186856-35186856 on Build GRCh38
rs750517053 Chr.11: 35186896-35186896 on Build GRCh38
rs758607544 Chr.11: 35186905-35186905 on Build GRCh38
rs1000780117 Chr.11: 35186813-35186813 on Build GRCh38
rs537853806 Chr.11: 35186935-35186935 on Build GRCh38
rs578210520 Chr.11: 35186961-35186961 on Build GRCh38
rs777131826 Chr.11: 35186933-35186933 on Build GRCh38
rs781643012 Chr.11: 35186918-35186918 on Build GRCh38
rs755248862 Chr.11: 35186914-35186914 on Build GRCh38
rs1031372965 Chr.11: 35186818-35186818 on Build GRCh38
rs35747418 Chr.11: 35186800-35186801 on Build GRCh38
rs756109723 Chr.11: 35186869-35186869 on Build GRCh38
rs1006700841 Chr.11: 35186787-35186787 on Build GRCh38
rs754328552 Chr.11: 35186934-35186934 on Build GRCh38
rs748711433 Chr.11: 35186797-35186797 on Build GRCh38
rs772778445 Chr.11: 35186802-35186802 on Build GRCh38
rs568808166 Chr.11: 35186923-35186923 on Build GRCh38
rs531113522 Chr.11: 35186819-35186820 on Build GRCh38
rs769142156 Chr.11: 35186799-35186799 on Build GRCh38

View Full Product Details