Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131199412-131199639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP7
Entrez Gene ID
9465
Gene Name
A-kinase anchoring protein 7
Gene Aliases
AKAP15, AKAP18
UniGene
Hs.486483
Gene Chromosome Location
Chr.6: 131125698-131283535 on Build GRCh38
Transcripts
NM_016377.3 XM_005267228.1 XM_005267229.4 XM_006715614.2 XM_011536262.1 XM_017011506.1 XM_017011507.1 XM_017011508.1 XM_017011509.1 XM_017011510.1 XM_017011511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5161657 c.636+5G>C p.? Chr.6: 131199578-131199578 on Build GRCh38
7237412 c.586C>T p.P196S Chr.6: 131199523-131199523 on Build GRCh38
4389678 c.630T>A p.R210R Chr.6: 131199567-131199567 on Build GRCh38
5404354 c.524-1G>A p.? Chr.6: 131199460-131199460 on Build GRCh38
3173052 c.617C>T p.S206L Chr.6: 131199554-131199554 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771833703 Chr.6: 131199591-131199591 on Build GRCh38
rs781501081 Chr.6: 131199461-131199461 on Build GRCh38
rs764753628 Chr.6: 131199545-131199545 on Build GRCh38
rs376062191 Chr.6: 131199491-131199491 on Build GRCh38
rs971561877 Chr.6: 131199459-131199459 on Build GRCh38
rs972190252 Chr.6: 131199470-131199470 on Build GRCh38
rs754636604 Chr.6: 131199571-131199571 on Build GRCh38
rs757221582 Chr.6: 131199613-131199613 on Build GRCh38
rs745682477 Chr.6: 131199586-131199586 on Build GRCh38
rs565029638 Chr.6: 131199509-131199509 on Build GRCh38
rs1190788 Chr.6: 131199515-131199515 on Build GRCh38
rs757493816 Chr.6: 131199572-131199572 on Build GRCh38
rs34324046 Chr.6: 131199551-131199551 on Build GRCh38
rs761354464 Chr.6: 131199608-131199608 on Build GRCh38
rs777870933 Chr.6: 131199565-131199565 on Build GRCh38
rs34221901 Chr.6: 131199502-131199502 on Build GRCh38
rs778947568 Chr.6: 131199581-131199581 on Build GRCh38
rs775345528 Chr.6: 131199592-131199592 on Build GRCh38
rs768678662 Chr.6: 131199601-131199601 on Build GRCh38
rs764537929 Chr.6: 131199612-131199612 on Build GRCh38
rs748272020 Chr.6: 131199467-131199467 on Build GRCh38
rs572683373 Chr.6: 131199487-131199487 on Build GRCh38
rs755537183 Chr.6: 131199450-131199450 on Build GRCh38
rs775226831 Chr.6: 131199507-131199507 on Build GRCh38
rs373824308 Chr.6: 131199550-131199550 on Build GRCh38
rs548274203 Chr.6: 131199614-131199614 on Build GRCh38
rs975714561 Chr.6: 131199557-131199557 on Build GRCh38
rs943501326 Chr.6: 131199533-131199533 on Build GRCh38
rs749562697 Chr.6: 131199483-131199483 on Build GRCh38
rs755238606 Chr.6: 131199548-131199548 on Build GRCh38
rs762662747 Chr.6: 131199610-131199610 on Build GRCh38
rs560762492 Chr.6: 131199444-131199444 on Build GRCh38
rs764954312 Chr.6: 131199609-131199609 on Build GRCh38
rs753373278 Chr.6: 131199522-131199522 on Build GRCh38
rs777825640 Chr.6: 131199482-131199482 on Build GRCh38
rs191862541 Chr.6: 131199597-131199597 on Build GRCh38
rs769797738 Chr.6: 131199475-131199475 on Build GRCh38
rs925473052 Chr.6: 131199529-131199529 on Build GRCh38
rs767828671 Chr.6: 131199611-131199611 on Build GRCh38
rs756591485 Chr.6: 131199558-131199558 on Build GRCh38
rs776309616 Chr.6: 131199606-131199606 on Build GRCh38
rs186732768 Chr.6: 131199566-131199566 on Build GRCh38
rs746757023 Chr.6: 131199618-131199618 on Build GRCh38
rs567893986 Chr.6: 131199616-131199616 on Build GRCh38
rs751973287 Chr.6: 131199547-131199547 on Build GRCh38

View Full Product Details