Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 68846170-68846409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC102B
Entrez Gene ID
79839
Gene Name
coiled-coil domain containing 102B
Gene Aliases
ACY1L, AN, C18orf14, HsT1731
UniGene
Hs.280781
Gene Chromosome Location
Chr.18: 68715221-69214350 on Build GRCh38
Transcripts
NM_001093729.1 NM_024781.2 XM_011526174.2 XM_011526175.2 XM_017025973.1 XM_017025974.1 XM_017025975.1 XM_017025976.1 XM_017025977.1 XM_017025978.1 XM_017025979.1 XM_017025980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7158044 c.901G>A p.E301K Chr.18: 68846386-68846386 on Build GRCh38
4381504 c.893A>G p.K298R Chr.18: 68846378-68846378 on Build GRCh38
4561974 c.906G>A p.L302L Chr.18: 68846391-68846391 on Build GRCh38
6276217 c.927G>T p.V309V Chr.18: 68846369-68846369 on Build GRCh38
148442 c.893A>G p.K298R Chr.18: 68846378-68846378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763923767 Chr.18: 68846310-68846310 on Build GRCh38
rs372574927 Chr.18: 68846354-68846354 on Build GRCh38
rs1954878 Chr.18: 68846234-68846234 on Build GRCh38
rs1040627920 Chr.18: 68846229-68846229 on Build GRCh38
rs772539222 Chr.18: 68846365-68846365 on Build GRCh38
rs186385550 Chr.18: 68846221-68846221 on Build GRCh38
rs759625130 Chr.18: 68846280-68846280 on Build GRCh38
rs778396864 Chr.18: 68846370-68846370 on Build GRCh38
rs534392919 Chr.18: 68846226-68846226 on Build GRCh38
rs2187094 Chr.18: 68846378-68846378 on Build GRCh38
rs762154731 Chr.18: 68846340-68846343 on Build GRCh38
rs183018793 Chr.18: 68846276-68846276 on Build GRCh38
rs368864406 Chr.18: 68846317-68846317 on Build GRCh38
rs765506898 Chr.18: 68846290-68846290 on Build GRCh38
rs267605245 Chr.18: 68846323-68846323 on Build GRCh38
rs752366694 Chr.18: 68846294-68846294 on Build GRCh38
rs747474605 Chr.18: 68846371-68846371 on Build GRCh38
rs771170694 Chr.18: 68846278-68846278 on Build GRCh38
rs186200166 Chr.18: 68846318-68846318 on Build GRCh38
rs140414838 Chr.18: 68846375-68846375 on Build GRCh38
rs776878874 Chr.18: 68846279-68846279 on Build GRCh38
rs376410494 Chr.18: 68846379-68846379 on Build GRCh38
rs762871870 Chr.18: 68846308-68846308 on Build GRCh38
rs542106970 Chr.18: 68846283-68846283 on Build GRCh38
rs190776188 Chr.18: 68846261-68846261 on Build GRCh38
rs147849810 Chr.18: 68846355-68846355 on Build GRCh38
rs774608767 Chr.18: 68846286-68846290 on Build GRCh38
rs913459541 Chr.18: 68846242-68846242 on Build GRCh38
rs749950683 Chr.18: 68846326-68846326 on Build GRCh38
rs191442848 Chr.18: 68846328-68846328 on Build GRCh38
rs771583661 Chr.18: 68846372-68846372 on Build GRCh38
rs980587688 Chr.18: 68846210-68846210 on Build GRCh38

View Full Product Details