Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119732087-119732344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAATS1
Entrez Gene ID
89876
Gene Name
MYCBP associated and testis expressed 1
Gene Aliases
AAT1, AAT1alpha, C3orf15, CaM-IP2, SPATA26
UniGene
Hs.341906
Gene Chromosome Location
Chr.3: 119703022-119767102 on Build GRCh38
Transcripts
NM_001320316.1 NM_001320317.1 NM_001320318.1 NM_033364.3 XM_011513277.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7239185 c.1051A>G p.I351V Chr.3: 119732326-119732326 on Build GRCh38
4667672 c.1068G>A p.E356E Chr.3: 119732343-119732343 on Build GRCh38
7304183 c.1067A>C p.E356A Chr.3: 119732342-119732342 on Build GRCh38
3845925 c.1047G>C p.K349N Chr.3: 119732322-119732322 on Build GRCh38
1036781 c.1043G>T p.R348I Chr.3: 119732318-119732318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74898621 Chr.3: 119732124-119732124 on Build GRCh38
rs368865309 Chr.3: 119732275-119732276 on Build GRCh38
rs932715292 Chr.3: 119732156-119732156 on Build GRCh38
rs767443871 Chr.3: 119732261-119732308 on Build GRCh38
rs759432638 Chr.3: 119732112-119732112 on Build GRCh38
rs765405230 Chr.3: 119732297-119732297 on Build GRCh38
rs560744648 Chr.3: 119732212-119732212 on Build GRCh38
rs774331598 Chr.3: 119732257-119732257 on Build GRCh38
rs746721615 Chr.3: 119732290-119732290 on Build GRCh38
rs761804420 Chr.3: 119732269-119732269 on Build GRCh38
rs919183048 Chr.3: 119732271-119732271 on Build GRCh38
rs888424914 Chr.3: 119732193-119732193 on Build GRCh38
rs375223778 Chr.3: 119732282-119732282 on Build GRCh38
rs564802939 Chr.3: 119732256-119732256 on Build GRCh38
rs575155318 Chr.3: 119732111-119732111 on Build GRCh38
rs760486175 Chr.3: 119732295-119732295 on Build GRCh38
rs768064623 Chr.3: 119732249-119732249 on Build GRCh38
rs1015935417 Chr.3: 119732209-119732209 on Build GRCh38
rs528200165 Chr.3: 119732218-119732218 on Build GRCh38
rs991191592 Chr.3: 119732165-119732165 on Build GRCh38
rs903406033 Chr.3: 119732118-119732118 on Build GRCh38
rs757507587 Chr.3: 119732301-119732302 on Build GRCh38
rs183293515 Chr.3: 119732224-119732224 on Build GRCh38
rs752482902 Chr.3: 119732273-119732274 on Build GRCh38
rs921382640 Chr.3: 119732128-119732128 on Build GRCh38
rs896061738 Chr.3: 119732234-119732234 on Build GRCh38
rs866376986 Chr.3: 119732302-119732302 on Build GRCh38
rs767267394 Chr.3: 119732273-119732273 on Build GRCh38
rs550386584 Chr.3: 119732222-119732222 on Build GRCh38
rs762454369 Chr.3: 119732245-119732245 on Build GRCh38
rs1035845761 Chr.3: 119732143-119732143 on Build GRCh38
rs905639219 Chr.3: 119732292-119732292 on Build GRCh38
rs958739554 Chr.3: 119732155-119732155 on Build GRCh38
rs752867506 Chr.3: 119732301-119732301 on Build GRCh38

View Full Product Details