Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 97581249-97581496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD2
Entrez Gene ID
26287
Gene Name
ankyrin repeat domain 2
Gene Aliases
ARPP
UniGene
Hs.73708
Gene Chromosome Location
Chr.10: 97572441-97583884 on Build GRCh38
Transcripts
NM_001129981.2 NM_001291218.1 NM_001291219.1 NM_020349.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4661792 c.670delG p.G225fs*3 Chr.10: 97581349-97581349 on Build GRCh38
4017082 c.735+1G>A p.? Chr.10: 97581415-97581415 on Build GRCh38
6303218 c.684G>T p.E228D Chr.10: 97581363-97581363 on Build GRCh38
5088169 c.640_643delGACT p.D214fs*13 Chr.10: 97581319-97581322 on Build GRCh38
6067073 c.660G>T p.W220C Chr.10: 97581339-97581339 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765002523 Chr.10: 97581422-97581422 on Build GRCh38
rs766215455 Chr.10: 97581321-97581321 on Build GRCh38
rs375975107 Chr.10: 97581360-97581360 on Build GRCh38
rs769145126 Chr.10: 97581364-97581364 on Build GRCh38
rs912099124 Chr.10: 97581449-97581449 on Build GRCh38
rs777018948 Chr.10: 97581366-97581366 on Build GRCh38
rs752356165 Chr.10: 97581421-97581422 on Build GRCh38
rs781193452 Chr.10: 97581342-97581342 on Build GRCh38
rs759095435 Chr.10: 97581416-97581416 on Build GRCh38
rs751642438 Chr.10: 97581427-97581427 on Build GRCh38
rs2275588 Chr.10: 97581428-97581428 on Build GRCh38
rs765413689 Chr.10: 97581277-97581277 on Build GRCh38
rs762088972 Chr.10: 97581273-97581273 on Build GRCh38
rs1004674541 Chr.10: 97581319-97581319 on Build GRCh38
rs375427070 Chr.10: 97581387-97581387 on Build GRCh38
rs766438735 Chr.10: 97581298-97581298 on Build GRCh38
rs773333184 Chr.10: 97581393-97581393 on Build GRCh38
rs758425825 Chr.10: 97581297-97581297 on Build GRCh38
rs150295757 Chr.10: 97581349-97581349 on Build GRCh38
rs766380384 Chr.10: 97581415-97581415 on Build GRCh38
rs763165659 Chr.10: 97581401-97581401 on Build GRCh38
rs141168302 Chr.10: 97581336-97581336 on Build GRCh38
rs747956625 Chr.10: 97581346-97581346 on Build GRCh38
rs759384092 Chr.10: 97581451-97581451 on Build GRCh38
rs189628043 Chr.10: 97581468-97581468 on Build GRCh38
rs755999979 Chr.10: 97581462-97581462 on Build GRCh38
rs545029249 Chr.10: 97581373-97581373 on Build GRCh38
rs760581249 Chr.10: 97581303-97581303 on Build GRCh38
rs767533923 Chr.10: 97581439-97581439 on Build GRCh38
rs373416347 Chr.10: 97581294-97581294 on Build GRCh38
rs779636010 Chr.10: 97581369-97581369 on Build GRCh38
rs747530603 Chr.10: 97581361-97581361 on Build GRCh38
rs559852218 Chr.10: 97581348-97581348 on Build GRCh38
rs1024719418 Chr.10: 97581343-97581343 on Build GRCh38
rs1002426643 Chr.10: 97581363-97581363 on Build GRCh38
rs531104474 Chr.10: 97581453-97581453 on Build GRCh38
rs867019853 Chr.10: 97581414-97581414 on Build GRCh38
rs770380531 Chr.10: 97581356-97581356 on Build GRCh38
rs113798764 Chr.10: 97581459-97581459 on Build GRCh38
rs751453940 Chr.10: 97581299-97581299 on Build GRCh38
rs200174082 Chr.10: 97581347-97581347 on Build GRCh38

View Full Product Details