Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101625486-101625757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX3
Entrez Gene ID
51566
Gene Name
armadillo repeat containing, X-linked 3
Gene Aliases
ALEX3, GASP6, dJ545K15.2
UniGene
Hs.592225
Gene Chromosome Location
Chr.X: 101623127-101631910 on Build GRCh38
Transcripts
NM_016607.3 NM_177947.2 NM_177948.2 XM_005262141.2

SNP Detail

SNP ID Chromosome Location
rs782745550 Chr.X: 101625678-101625678 on Build GRCh38
rs782000125 Chr.X: 101625648-101625648 on Build GRCh38
rs782461535 Chr.X: 101625688-101625688 on Build GRCh38
rs376915704 Chr.X: 101625732-101625732 on Build GRCh38
rs782302901 Chr.X: 101625675-101625675 on Build GRCh38
rs781882226 Chr.X: 101625712-101625712 on Build GRCh38
rs372051555 Chr.X: 101625571-101625571 on Build GRCh38
rs782505708 Chr.X: 101625720-101625720 on Build GRCh38
rs997449630 Chr.X: 101625715-101625715 on Build GRCh38
rs782809337 Chr.X: 101625585-101625585 on Build GRCh38
rs782809877 Chr.X: 101625695-101625695 on Build GRCh38
rs781828907 Chr.X: 101625680-101625680 on Build GRCh38
rs781926229 Chr.X: 101625561-101625561 on Build GRCh38

View Full Product Details