Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 69386890-69387159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERH
Entrez Gene ID
2079
Gene Name
enhancer of rudimentary homolog (Drosophila)
Gene Aliases
DROER
UniGene
Hs.509791
Gene Chromosome Location
Chr.14: 69380123-69398304 on Build GRCh38
Transcripts
NM_004450.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7154770 c.201C>T p.L67L Chr.14: 69386974-69386974 on Build GRCh38
5946353 c.212+2T>G p.? Chr.14: 69386961-69386961 on Build GRCh38
6993373 c.146C>T p.S49F Chr.14: 69387029-69387029 on Build GRCh38
2135613 c.163A>G p.S55G Chr.14: 69387012-69387012 on Build GRCh38
6521620 c.105G>T p.M35I Chr.14: 69387070-69387070 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746510288 Chr.14: 69387129-69387129 on Build GRCh38
rs776318855 Chr.14: 69386921-69386921 on Build GRCh38
rs1007655477 Chr.14: 69387134-69387134 on Build GRCh38
rs777103056 Chr.14: 69387122-69387122 on Build GRCh38
rs774519236 Chr.14: 69386992-69386992 on Build GRCh38
rs995559821 Chr.14: 69387004-69387004 on Build GRCh38
rs762077712 Chr.14: 69387007-69387007 on Build GRCh38
rs116464054 Chr.14: 69387105-69387105 on Build GRCh38
rs762963699 Chr.14: 69386965-69386965 on Build GRCh38
rs112914904 Chr.14: 69387086-69387086 on Build GRCh38
rs767825914 Chr.14: 69387095-69387095 on Build GRCh38
rs139950170 Chr.14: 69386980-69386980 on Build GRCh38
rs370350818 Chr.14: 69387121-69387121 on Build GRCh38
rs769644674 Chr.14: 69386952-69386952 on Build GRCh38
rs200718090 Chr.14: 69387112-69387112 on Build GRCh38
rs745632194 Chr.14: 69386926-69386926 on Build GRCh38
rs750710139 Chr.14: 69387099-69387099 on Build GRCh38
rs939250891 Chr.14: 69386977-69386977 on Build GRCh38
rs1048827682 Chr.14: 69387127-69387127 on Build GRCh38
rs775278303 Chr.14: 69386957-69386957 on Build GRCh38
rs897657756 Chr.14: 69387092-69387092 on Build GRCh38

View Full Product Details