Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 19906092-19906365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP10
Entrez Gene ID
11216
Gene Name
A-kinase anchoring protein 10
Gene Aliases
AKAP-10, D-AKAP-2, D-AKAP2, PRKA10
UniGene
Hs.642676
Gene Chromosome Location
Chr.17: 19904452-19977856 on Build GRCh38
Transcripts
NM_007202.3 XM_006721432.3

SNP Detail

SNP ID Chromosome Location
rs759870915 Chr.17: 19906178-19906178 on Build GRCh38
rs771090509 Chr.17: 19906278-19906278 on Build GRCh38
rs754704606 Chr.17: 19906226-19906226 on Build GRCh38
rs534145009 Chr.17: 19906172-19906172 on Build GRCh38
rs910690507 Chr.17: 19906145-19906145 on Build GRCh38
rs749889990 Chr.17: 19906202-19906202 on Build GRCh38
rs761950011 Chr.17: 19906120-19906120 on Build GRCh38
rs999138066 Chr.17: 19906117-19906117 on Build GRCh38
rs376371619 Chr.17: 19906203-19906203 on Build GRCh38
rs747967068 Chr.17: 19906242-19906242 on Build GRCh38
rs780025259 Chr.17: 19906338-19906338 on Build GRCh38
rs780758449 Chr.17: 19906235-19906236 on Build GRCh38
rs923510717 Chr.17: 19906194-19906194 on Build GRCh38
rs995395540 Chr.17: 19906274-19906274 on Build GRCh38
rs764676780 Chr.17: 19906198-19906198 on Build GRCh38
rs756354902 Chr.17: 19906296-19906296 on Build GRCh38
rs371785532 Chr.17: 19906195-19906195 on Build GRCh38
rs758251704 Chr.17: 19906247-19906247 on Build GRCh38
rs753598190 Chr.17: 19906224-19906224 on Build GRCh38
rs777828938 Chr.17: 19906254-19906254 on Build GRCh38
rs763301253 Chr.17: 19906190-19906190 on Build GRCh38
rs374260696 Chr.17: 19906339-19906339 on Build GRCh38
rs747572284 Chr.17: 19906238-19906242 on Build GRCh38
rs769509545 Chr.17: 19906237-19906238 on Build GRCh38
rs370871830 Chr.17: 19906130-19906130 on Build GRCh38
rs896185564 Chr.17: 19906154-19906154 on Build GRCh38
rs778813662 Chr.17: 19906230-19906230 on Build GRCh38
rs76583306 Chr.17: 19906138-19906138 on Build GRCh38
rs976362291 Chr.17: 19906179-19906179 on Build GRCh38
rs553914095 Chr.17: 19906275-19906275 on Build GRCh38
rs935122234 Chr.17: 19906131-19906131 on Build GRCh38
rs766099873 Chr.17: 19906205-19906205 on Build GRCh38
rs750685621 Chr.17: 19906151-19906151 on Build GRCh38
rs765536820 Chr.17: 19906183-19906183 on Build GRCh38
rs775862047 Chr.17: 19906187-19906187 on Build GRCh38
rs184159200 Chr.17: 19906135-19906135 on Build GRCh38

View Full Product Details