Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73554924-73555149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRA2
Entrez Gene ID
57763
Gene Name
ankyrin repeat family A member 2
Gene Aliases
ANKRA
UniGene
Hs.239154
Gene Chromosome Location
Chr.5: 73552200-73565705 on Build GRCh38
Transcripts
NM_023039.4 XM_005248560.2 XM_017009678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5501878 c.641G>T p.G214V Chr.5: 73554958-73554958 on Build GRCh38
3661936 c.666C>T p.A222A Chr.5: 73554933-73554933 on Build GRCh38
4661726 c.660G>A p.S220S Chr.5: 73554939-73554939 on Build GRCh38
5061189 c.622C>A p.P208T Chr.5: 73554977-73554977 on Build GRCh38
1438510 c.643C>T p.R215* Chr.5: 73554956-73554956 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs988142764 Chr.5: 73554958-73554958 on Build GRCh38
rs768294997 Chr.5: 73554967-73554967 on Build GRCh38
rs1043372157 Chr.5: 73554996-73554996 on Build GRCh38
rs991275847 Chr.5: 73555099-73555099 on Build GRCh38
rs776804845 Chr.5: 73555022-73555022 on Build GRCh38
rs1020632424 Chr.5: 73555018-73555018 on Build GRCh38
rs761659654 Chr.5: 73555036-73555036 on Build GRCh38
rs748540090 Chr.5: 73554995-73554995 on Build GRCh38
rs779148333 Chr.5: 73554993-73554993 on Build GRCh38
rs150789725 Chr.5: 73554956-73554956 on Build GRCh38
rs192928803 Chr.5: 73555051-73555051 on Build GRCh38
rs748959327 Chr.5: 73554960-73554960 on Build GRCh38
rs760877706 Chr.5: 73555012-73555012 on Build GRCh38
rs773746633 Chr.5: 73555010-73555010 on Build GRCh38
rs981272933 Chr.5: 73555057-73555057 on Build GRCh38
rs771166593 Chr.5: 73555020-73555020 on Build GRCh38
rs967263169 Chr.5: 73555053-73555053 on Build GRCh38
rs759853416 Chr.5: 73555023-73555023 on Build GRCh38
rs931000981 Chr.5: 73555111-73555111 on Build GRCh38
rs563334606 Chr.5: 73555082-73555082 on Build GRCh38
rs928416326 Chr.5: 73555064-73555064 on Build GRCh38
rs777019325 Chr.5: 73555027-73555028 on Build GRCh38
rs751380541 Chr.5: 73555034-73555034 on Build GRCh38
rs958519704 Chr.5: 73555068-73555068 on Build GRCh38
rs772511692 Chr.5: 73555000-73555000 on Build GRCh38
rs879713427 Chr.5: 73555101-73555101 on Build GRCh38
rs903815596 Chr.5: 73555029-73555029 on Build GRCh38
rs765436428 Chr.5: 73555031-73555031 on Build GRCh38
rs1002086086 Chr.5: 73555042-73555042 on Build GRCh38
rs577768694 Chr.5: 73554955-73554955 on Build GRCh38

View Full Product Details