Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 45157734-45157969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXorf36
Entrez Gene ID
79742
Gene Name
chromosome X open reading frame 36
Gene Aliases
4930578C19Rik, DIA1R, EPQL1862, PRO3743, bA435K1.1
UniGene
Hs.98321
Gene Chromosome Location
Chr.X: 45148373-45200901 on Build GRCh38
Transcripts
NM_176819.3 XM_005272670.1 XM_006724559.1

SNP Detail

SNP ID Chromosome Location
rs776800924 Chr.X: 45157801-45157801 on Build GRCh38
rs898728182 Chr.X: 45157915-45157915 on Build GRCh38
rs766133952 Chr.X: 45157824-45157824 on Build GRCh38
rs781118982 Chr.X: 45157923-45157924 on Build GRCh38
rs780524532 Chr.X: 45157911-45157911 on Build GRCh38
rs773525670 Chr.X: 45157929-45157929 on Build GRCh38
rs755578524 Chr.X: 45157866-45157866 on Build GRCh38
rs764113239 Chr.X: 45157846-45157846 on Build GRCh38
rs774573891 Chr.X: 45157856-45157856 on Build GRCh38
rs916240409 Chr.X: 45157811-45157811 on Build GRCh38
rs993805247 Chr.X: 45157792-45157792 on Build GRCh38
rs777536372 Chr.X: 45157874-45157874 on Build GRCh38
rs773294215 Chr.X: 45157808-45157808 on Build GRCh38
rs752189433 Chr.X: 45157865-45157865 on Build GRCh38
rs887602649 Chr.X: 45157857-45157857 on Build GRCh38
rs748319047 Chr.X: 45157926-45157926 on Build GRCh38
rs949042612 Chr.X: 45157915-45157917 on Build GRCh38
rs746307645 Chr.X: 45157761-45157761 on Build GRCh38
rs5952693 Chr.X: 45157921-45157921 on Build GRCh38
rs769739911 Chr.X: 45157927-45157927 on Build GRCh38
rs780753232 Chr.X: 45157778-45157778 on Build GRCh38
rs747539687 Chr.X: 45157912-45157912 on Build GRCh38
rs937688398 Chr.X: 45157760-45157760 on Build GRCh38
rs775676593 Chr.X: 45157765-45157765 on Build GRCh38
rs61423499 Chr.X: 45157946-45157946 on Build GRCh38
rs769990784 Chr.X: 45157806-45157806 on Build GRCh38
rs747484633 Chr.X: 45157786-45157786 on Build GRCh38
rs1017034992 Chr.X: 45157894-45157894 on Build GRCh38
rs751530771 Chr.X: 45157831-45157831 on Build GRCh38
rs990579865 Chr.X: 45157773-45157773 on Build GRCh38
rs757093065 Chr.X: 45157774-45157774 on Build GRCh38
rs1047588819 Chr.X: 45157807-45157807 on Build GRCh38
rs761954583 Chr.X: 45157805-45157805 on Build GRCh38
rs374071629 Chr.X: 45157886-45157886 on Build GRCh38
rs371914100 Chr.X: 45157771-45157771 on Build GRCh38
rs1005681023 Chr.X: 45157875-45157875 on Build GRCh38
rs758871668 Chr.X: 45157892-45157892 on Build GRCh38
rs768886940 Chr.X: 45157790-45157790 on Build GRCh38
rs111962847 Chr.X: 45157876-45157876 on Build GRCh38
rs763055334 Chr.X: 45157820-45157820 on Build GRCh38

View Full Product Details