Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 131535232-131535459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE4
Entrez Gene ID
131034
Gene Name
copine 4
Gene Aliases
COPN4, CPN4
UniGene
Hs.199877
Gene Chromosome Location
Chr.3: 131533560-132285702 on Build GRCh38
Transcripts
NM_001289112.1 NM_130808.2 NM_153429.1 XM_011512406.2 XM_011512407.2 XM_011512408.2 XM_017005693.1 XM_017005695.1 XM_017005696.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5418564 c.1606G>A p.A536T Chr.3: 131535317-131535317 on Build GRCh38
6252431 c.1620C>A p.S540R Chr.3: 131535303-131535303 on Build GRCh38
3992736 c.1541C>T p.A514V Chr.3: 131535328-131535328 on Build GRCh38
4674376 c.1670G>A p.G557E Chr.3: 131535253-131535253 on Build GRCh38
5625281 c.1653C>T p.D551D Chr.3: 131535270-131535270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772404448 Chr.3: 131535381-131535381 on Build GRCh38
rs758131519 Chr.3: 131535333-131535333 on Build GRCh38
rs561689852 Chr.3: 131535304-131535304 on Build GRCh38
rs913673496 Chr.3: 131535260-131535260 on Build GRCh38
rs76599136 Chr.3: 131535386-131535386 on Build GRCh38
rs774657683 Chr.3: 131535270-131535270 on Build GRCh38
rs765129150 Chr.3: 131535327-131535327 on Build GRCh38
rs779343771 Chr.3: 131535368-131535368 on Build GRCh38
rs777289459 Chr.3: 131535334-131535334 on Build GRCh38
rs987130076 Chr.3: 131535329-131535329 on Build GRCh38
rs762098925 Chr.3: 131535273-131535273 on Build GRCh38
rs920396529 Chr.3: 131535421-131535421 on Build GRCh38
rs533532161 Chr.3: 131535428-131535428 on Build GRCh38
rs193182961 Chr.3: 131535356-131535356 on Build GRCh38
rs759507626 Chr.3: 131535317-131535317 on Build GRCh38
rs780629970 Chr.3: 131535358-131535358 on Build GRCh38
rs527585476 Chr.3: 131535357-131535357 on Build GRCh38
rs373539566 Chr.3: 131535367-131535367 on Build GRCh38
rs372459106 Chr.3: 131535267-131535267 on Build GRCh38
rs748447091 Chr.3: 131535370-131535370 on Build GRCh38
rs772123965 Chr.3: 131535286-131535286 on Build GRCh38
rs760663500 Chr.3: 131535303-131535303 on Build GRCh38
rs773395113 Chr.3: 131535302-131535302 on Build GRCh38
rs753851188 Chr.3: 131535311-131535311 on Build GRCh38
rs370057154 Chr.3: 131535362-131535362 on Build GRCh38
rs923521093 Chr.3: 131535394-131535394 on Build GRCh38
rs751124415 Chr.3: 131535341-131535341 on Build GRCh38

View Full Product Details