Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128480004-128480277 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETS1
Entrez Gene ID
2113
Gene Name
ETS proto-oncogene 1, transcription factor
Gene Aliases
ETS-1, EWSR2, c-ets-1, p54
UniGene
Hs.369438
Gene Chromosome Location
Chr.11: 128458761-128587593 on Build GRCh38
Transcripts
NM_001143820.1 NM_001162422.1 NM_005238.3 XM_011542649.2 XM_011542650.1 XM_011542651.1 XM_011542652.1 XM_017017314.1 XM_017017315.1 XM_017017316.1 XM_017017317.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4019242 c.1063C>T p.R355C Chr.11: 128480251-128480251 on Build GRCh38
5372004 c.940C>T p.L314F Chr.11: 128480242-128480242 on Build GRCh38
6335047 c.1063C>A p.R355S Chr.11: 128480251-128480251 on Build GRCh38
5372003 c.1072C>T p.L358F Chr.11: 128480242-128480242 on Build GRCh38
4019243 c.931C>T p.R311C Chr.11: 128480251-128480251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748395139 Chr.11: 128480230-128480230 on Build GRCh38
rs757231712 Chr.11: 128480183-128480183 on Build GRCh38
rs568554221 Chr.11: 128480044-128480044 on Build GRCh38
rs570350607 Chr.11: 128480177-128480177 on Build GRCh38
rs774134665 Chr.11: 128480153-128480153 on Build GRCh38
rs1002283773 Chr.11: 128480097-128480097 on Build GRCh38
rs752364345 Chr.11: 128480169-128480169 on Build GRCh38
rs1020796728 Chr.11: 128480124-128480124 on Build GRCh38
rs149270563 Chr.11: 128480151-128480151 on Build GRCh38
rs997937653 Chr.11: 128480101-128480101 on Build GRCh38
rs1031802289 Chr.11: 128480035-128480035 on Build GRCh38
rs778702464 Chr.11: 128480184-128480184 on Build GRCh38
rs768693175 Chr.11: 128480187-128480187 on Build GRCh38
rs548010220 Chr.11: 128480074-128480074 on Build GRCh38
rs756020577 Chr.11: 128480175-128480175 on Build GRCh38
rs201962711 Chr.11: 128480185-128480185 on Build GRCh38
rs953675364 Chr.11: 128480110-128480110 on Build GRCh38
rs533058737 Chr.11: 128480207-128480207 on Build GRCh38
rs1034217422 Chr.11: 128480098-128480098 on Build GRCh38
rs61909072 Chr.11: 128480176-128480176 on Build GRCh38
rs767343500 Chr.11: 128480162-128480162 on Build GRCh38
rs755152863 Chr.11: 128480140-128480140 on Build GRCh38
rs1029364522 Chr.11: 128480104-128480104 on Build GRCh38
rs917270554 Chr.11: 128480250-128480250 on Build GRCh38
rs113082559 Chr.11: 128480189-128480189 on Build GRCh38
rs571000924 Chr.11: 128480082-128480082 on Build GRCh38
rs113375412 Chr.11: 128480163-128480163 on Build GRCh38
rs869218744 Chr.11: 128480086-128480086 on Build GRCh38
rs112740933 Chr.11: 128480073-128480073 on Build GRCh38
rs769966830 Chr.11: 128480231-128480231 on Build GRCh38
rs766964257 Chr.11: 128480257-128480257 on Build GRCh38
rs763167051 Chr.11: 128480251-128480251 on Build GRCh38
rs957876161 Chr.11: 128480107-128480107 on Build GRCh38
rs1016873341 Chr.11: 128480156-128480156 on Build GRCh38
rs773626156 Chr.11: 128480238-128480238 on Build GRCh38
rs985100037 Chr.11: 128480138-128480138 on Build GRCh38

View Full Product Details