Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11381509-11381770 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPOR
Entrez Gene ID
2057
Gene Name
erythropoietin receptor
Gene Aliases
EPO-R
UniGene
Hs.631624
Gene Chromosome Location
Chr.19: 11377205-11384342 on Build GRCh38
Transcripts
NM_000121.3 NR_033663.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4616507 c.559G>C p.G187R Chr.19: 11381718-11381718 on Build GRCh38
7329715 c.529C>G p.H177D Chr.19: 11381748-11381748 on Build GRCh38
5530581 c.534C>A p.I178I Chr.19: 11381743-11381743 on Build GRCh38
7242868 c.541G>T p.E181* Chr.19: 11381736-11381736 on Build GRCh38
4833142 c.529C>T p.H177Y Chr.19: 11381748-11381748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770002611 Chr.19: 11381646-11381646 on Build GRCh38
rs756727018 Chr.19: 11381750-11381750 on Build GRCh38
rs778494949 Chr.19: 11381692-11381692 on Build GRCh38
rs1048974599 Chr.19: 11381546-11381546 on Build GRCh38
rs377285358 Chr.19: 11381660-11381660 on Build GRCh38
rs562676037 Chr.19: 11381554-11381554 on Build GRCh38
rs775215856 Chr.19: 11381718-11381718 on Build GRCh38
rs770377268 Chr.19: 11381699-11381699 on Build GRCh38
rs748682367 Chr.19: 11381698-11381698 on Build GRCh38
rs758069237 Chr.19: 11381741-11381741 on Build GRCh38
rs369880281 Chr.19: 11381661-11381661 on Build GRCh38
rs748929970 Chr.19: 11381653-11381653 on Build GRCh38
rs1038487205 Chr.19: 11381639-11381639 on Build GRCh38
rs911266659 Chr.19: 11381622-11381622 on Build GRCh38
rs762053489 Chr.19: 11381645-11381645 on Build GRCh38
rs759686842 Chr.19: 11381713-11381713 on Build GRCh38
rs755986351 Chr.19: 11381665-11381665 on Build GRCh38
rs567166322 Chr.19: 11381686-11381686 on Build GRCh38
rs750107289 Chr.19: 11381737-11381737 on Build GRCh38
rs765009836 Chr.19: 11381736-11381736 on Build GRCh38
rs771235530 Chr.19: 11381711-11381711 on Build GRCh38
rs371950695 Chr.19: 11381647-11381647 on Build GRCh38
rs778403971 Chr.19: 11381680-11381680 on Build GRCh38
rs574619833 Chr.19: 11381670-11381670 on Build GRCh38
rs781710022 Chr.19: 11381744-11381744 on Build GRCh38
rs748484141 Chr.19: 11381746-11381746 on Build GRCh38
rs776503090 Chr.19: 11381662-11381663 on Build GRCh38
rs373277892 Chr.19: 11381674-11381674 on Build GRCh38
rs187912731 Chr.19: 11381556-11381556 on Build GRCh38
rs751412892 Chr.19: 11381659-11381659 on Build GRCh38
rs922886026 Chr.19: 11381540-11381540 on Build GRCh38
rs747467034 Chr.19: 11381684-11381684 on Build GRCh38
rs551636869 Chr.19: 11381632-11381632 on Build GRCh38
rs202198017 Chr.19: 11381664-11381664 on Build GRCh38
rs978732810 Chr.19: 11381539-11381539 on Build GRCh38
rs933966160 Chr.19: 11381604-11381604 on Build GRCh38
rs930064878 Chr.19: 11381533-11381533 on Build GRCh38
rs766471919 Chr.19: 11381656-11381656 on Build GRCh38
rs533141447 Chr.19: 11381596-11381596 on Build GRCh38
rs756706572 Chr.19: 11381722-11381722 on Build GRCh38
rs780039428 Chr.19: 11381610-11381610 on Build GRCh38
rs769140416 Chr.19: 11381689-11381689 on Build GRCh38
rs774430628 Chr.19: 11381712-11381712 on Build GRCh38
rs773372608 Chr.19: 11381701-11381701 on Build GRCh38
rs756971485 Chr.19: 11381675-11381675 on Build GRCh38
rs139849355 Chr.19: 11381709-11381709 on Build GRCh38
rs1054916047 Chr.19: 11381619-11381619 on Build GRCh38
rs887577521 Chr.19: 11381547-11381547 on Build GRCh38
rs377322757 Chr.19: 11381719-11381719 on Build GRCh38

View Full Product Details