Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 124670091-124670336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD50
Entrez Gene ID
57182
Gene Name
ankyrin repeat domain 50
Gene Aliases
-
UniGene
Hs.480694
Gene Chromosome Location
Chr.4: 124664048-124712732 on Build GRCh38
Transcripts
NM_001167882.1 NM_020337.2 XM_017008471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3009072 c.2965C>T p.Q989* Chr.4: 124670312-124670312 on Build GRCh38
4121891 c.3173T>A p.I1058N Chr.4: 124670104-124670104 on Build GRCh38
4121892 c.3033A>G p.E1011E Chr.4: 124670244-124670244 on Build GRCh38
4337604 c.3163delG p.E1055fs*11 Chr.4: 124670114-124670114 on Build GRCh38
5046049 c.3184C>G p.Q1062E Chr.4: 124670093-124670093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760903897 Chr.4: 124670310-124670310 on Build GRCh38
rs757664338 Chr.4: 124670145-124670145 on Build GRCh38
rs139499397 Chr.4: 124670299-124670299 on Build GRCh38
rs773106734 Chr.4: 124670170-124670170 on Build GRCh38
rs764738403 Chr.4: 124670227-124670227 on Build GRCh38
rs911364713 Chr.4: 124670146-124670146 on Build GRCh38
rs774453017 Chr.4: 124670176-124670176 on Build GRCh38
rs779918695 Chr.4: 124670151-124670151 on Build GRCh38
rs775586706 Chr.4: 124670208-124670208 on Build GRCh38
rs770702877 Chr.4: 124670174-124670174 on Build GRCh38
rs753145762 Chr.4: 124670119-124670119 on Build GRCh38
rs775544087 Chr.4: 124670308-124670308 on Build GRCh38
rs745819135 Chr.4: 124670262-124670262 on Build GRCh38
rs200351787 Chr.4: 124670130-124670130 on Build GRCh38
rs944175738 Chr.4: 124670182-124670182 on Build GRCh38
rs141095097 Chr.4: 124670265-124670265 on Build GRCh38
rs748058628 Chr.4: 124670162-124670162 on Build GRCh38
rs371573842 Chr.4: 124670273-124670273 on Build GRCh38
rs148154149 Chr.4: 124670260-124670260 on Build GRCh38
rs746096555 Chr.4: 124670303-124670303 on Build GRCh38
rs746657581 Chr.4: 124670154-124670154 on Build GRCh38
rs757069076 Chr.4: 124670202-124670202 on Build GRCh38
rs991406786 Chr.4: 124670309-124670309 on Build GRCh38
rs765633282 Chr.4: 124670239-124670239 on Build GRCh38
rs145805142 Chr.4: 124670240-124670240 on Build GRCh38
rs3822116 Chr.4: 124670268-124670268 on Build GRCh38
rs754303830 Chr.4: 124670144-124670144 on Build GRCh38
rs772382179 Chr.4: 124670305-124670305 on Build GRCh38
rs754611161 Chr.4: 124670244-124670244 on Build GRCh38
rs769772793 Chr.4: 124670168-124670168 on Build GRCh38
rs146823198 Chr.4: 124670171-124670171 on Build GRCh38
rs140645873 Chr.4: 124670190-124670190 on Build GRCh38
rs756600446 Chr.4: 124670123-124670123 on Build GRCh38
rs780715353 Chr.4: 124670157-124670157 on Build GRCh38
rs754391701 Chr.4: 124670229-124670229 on Build GRCh38
rs757752297 Chr.4: 124670233-124670233 on Build GRCh38
rs767522480 Chr.4: 124670199-124670199 on Build GRCh38

View Full Product Details