Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 150477034-150477303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIMAP8
Entrez Gene ID
155038
Gene Name
GTPase, IMAP family member 8
Gene Aliases
IAN-9, IAN6, IAN9, IANT
UniGene
Hs.647121
Gene Chromosome Location
Chr.7: 150450630-150479395 on Build GRCh38
Transcripts
NM_175571.3 XM_005249950.4 XM_005249951.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6276761 c.1471G>T p.V491F Chr.7: 150477253-150477253 on Build GRCh38
6238974 c.1358C>A p.A453E Chr.7: 150477140-150477140 on Build GRCh38
3923079 c.1421C>T p.T474I Chr.7: 150477203-150477203 on Build GRCh38
5626319 c.1465G>T p.V489L Chr.7: 150477247-150477247 on Build GRCh38
485039 c.1374C>T p.I458I Chr.7: 150477156-150477156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023070904 Chr.7: 150477082-150477082 on Build GRCh38
rs759052186 Chr.7: 150477092-150477092 on Build GRCh38
rs764873030 Chr.7: 150477104-150477104 on Build GRCh38
rs777656666 Chr.7: 150477221-150477221 on Build GRCh38
rs759939012 Chr.7: 150477157-150477157 on Build GRCh38
rs376371559 Chr.7: 150477141-150477141 on Build GRCh38
rs773835288 Chr.7: 150477070-150477070 on Build GRCh38
rs768306288 Chr.7: 150477246-150477246 on Build GRCh38
rs769600750 Chr.7: 150477237-150477251 on Build GRCh38
rs763552360 Chr.7: 150477168-150477168 on Build GRCh38
rs747031875 Chr.7: 150477079-150477079 on Build GRCh38
rs199561369 Chr.7: 150477238-150477238 on Build GRCh38
rs748341275 Chr.7: 150477188-150477188 on Build GRCh38
rs772247999 Chr.7: 150477144-150477144 on Build GRCh38
rs199866281 Chr.7: 150477226-150477226 on Build GRCh38
rs866619157 Chr.7: 150477277-150477277 on Build GRCh38
rs761285793 Chr.7: 150477081-150477081 on Build GRCh38
rs753699589 Chr.7: 150477269-150477269 on Build GRCh38
rs758626621 Chr.7: 150477279-150477279 on Build GRCh38
rs762631650 Chr.7: 150477166-150477166 on Build GRCh38
rs775099199 Chr.7: 150477165-150477165 on Build GRCh38
rs777075844 Chr.7: 150477089-150477089 on Build GRCh38
rs758716528 Chr.7: 150477190-150477190 on Build GRCh38
rs565000585 Chr.7: 150477135-150477135 on Build GRCh38
rs773372677 Chr.7: 150477145-150477145 on Build GRCh38
rs774509839 Chr.7: 150477061-150477061 on Build GRCh38
rs150230235 Chr.7: 150477186-150477186 on Build GRCh38
rs760726436 Chr.7: 150477260-150477260 on Build GRCh38
rs762381752 Chr.7: 150477265-150477265 on Build GRCh38
rs1014965282 Chr.7: 150477085-150477085 on Build GRCh38
rs745609301 Chr.7: 150477232-150477235 on Build GRCh38
rs200600015 Chr.7: 150477172-150477172 on Build GRCh38
rs200346050 Chr.7: 150477140-150477140 on Build GRCh38
rs200837107 Chr.7: 150477163-150477163 on Build GRCh38
rs750388336 Chr.7: 150477182-150477182 on Build GRCh38
rs770110198 Chr.7: 150477236-150477236 on Build GRCh38
rs777180069 Chr.7: 150477229-150477229 on Build GRCh38
rs747115766 Chr.7: 150477222-150477222 on Build GRCh38
rs912929423 Chr.7: 150477246-150477246 on Build GRCh38
rs773946101 Chr.7: 150477248-150477248 on Build GRCh38
rs766987168 Chr.7: 150477254-150477254 on Build GRCh38
rs775140418 Chr.7: 150477263-150477264 on Build GRCh38
rs111494335 Chr.7: 150477065-150477065 on Build GRCh38
rs752410990 Chr.7: 150477114-150477114 on Build GRCh38
rs201799511 Chr.7: 150477131-150477131 on Build GRCh38
rs529904898 Chr.7: 150477187-150477187 on Build GRCh38
rs757245861 Chr.7: 150477123-150477123 on Build GRCh38
rs372916812 Chr.7: 150477124-150477124 on Build GRCh38
rs945698342 Chr.7: 150477278-150477278 on Build GRCh38
rs751098205 Chr.7: 150477169-150477169 on Build GRCh38
rs775860008 Chr.7: 150477237-150477237 on Build GRCh38
rs745684371 Chr.7: 150477126-150477126 on Build GRCh38
rs750344929 Chr.7: 150477257-150477257 on Build GRCh38
rs138720266 Chr.7: 150477266-150477266 on Build GRCh38
rs368806122 Chr.7: 150477154-150477154 on Build GRCh38
rs757888592 Chr.7: 150477115-150477115 on Build GRCh38
rs201505121 Chr.7: 150477181-150477181 on Build GRCh38
rs895865579 Chr.7: 150477079-150477079 on Build GRCh38
rs763640048 Chr.7: 150477119-150477119 on Build GRCh38
rs751417524 Chr.7: 150477122-150477122 on Build GRCh38
rs140969132 Chr.7: 150477150-150477150 on Build GRCh38
rs746420800 Chr.7: 150477232-150477232 on Build GRCh38
rs781177204 Chr.7: 150477229-150477232 on Build GRCh38
rs761375805 Chr.7: 150477251-150477251 on Build GRCh38
rs770957794 Chr.7: 150477114-150477114 on Build GRCh38
rs766613797 Chr.7: 150477083-150477083 on Build GRCh38
rs866321472 Chr.7: 150477147-150477147 on Build GRCh38
rs772337910 Chr.7: 150477066-150477066 on Build GRCh38

View Full Product Details