Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3728232-3728505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBBP
Entrez Gene ID
1387
Gene Name
CREB binding protein
Gene Aliases
CBP, KAT3A, RSTS
UniGene
Hs.459759
Gene Chromosome Location
Chr.16: 3725054-3880727 on Build GRCh38
Transcripts
NM_001079846.1 NM_004380.2 XM_005255124.4 XM_005255125.4 XM_006720848.3 XM_011522381.2 XM_017022944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1202269 c.6596A>T p.Q2199L Chr.16: 3728451-3728451 on Build GRCh38
6914734 c.6609_6611delACA p.Q2216delQ Chr.16: 3728436-3728438 on Build GRCh38
1249111 c.6727G>A p.A2243T Chr.16: 3728320-3728320 on Build GRCh38
6498602 c.6745_6746insAGCAGC p.Q2248_R2249insQQ Chr.16: 3728301-3728302 on Build GRCh38
6848053 c.6772G>A p.G2258S Chr.16: 3728275-3728275 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767903262 Chr.16: 3728288-3728289 on Build GRCh38
rs778507028 Chr.16: 3728403-3728426 on Build GRCh38
rs570415368 Chr.16: 3728444-3728444 on Build GRCh38
rs763255288 Chr.16: 3728466-3728466 on Build GRCh38
rs752887380 Chr.16: 3728281-3728281 on Build GRCh38
rs903906594 Chr.16: 3728435-3728435 on Build GRCh38
rs138613594 Chr.16: 3728363-3728363 on Build GRCh38
rs746121736 Chr.16: 3728439-3728450 on Build GRCh38
rs190153828 Chr.16: 3728319-3728319 on Build GRCh38
rs778220523 Chr.16: 3728370-3728381 on Build GRCh38
rs778678603 Chr.16: 3728424-3728435 on Build GRCh38
rs868434987 Chr.16: 3728296-3728296 on Build GRCh38
rs764545471 Chr.16: 3728282-3728282 on Build GRCh38
rs775677510 Chr.16: 3728351-3728351 on Build GRCh38
rs754959530 Chr.16: 3728437-3728438 on Build GRCh38
rs776123525 Chr.16: 3728251-3728251 on Build GRCh38
rs779834061 Chr.16: 3728405-3728406 on Build GRCh38
rs753500354 Chr.16: 3728399-3728400 on Build GRCh38
rs200716582 Chr.16: 3728386-3728386 on Build GRCh38
rs765821935 Chr.16: 3728451-3728451 on Build GRCh38
rs141184151 Chr.16: 3728389-3728389 on Build GRCh38
rs766219870 Chr.16: 3728263-3728263 on Build GRCh38
rs760793692 Chr.16: 3728354-3728354 on Build GRCh38
rs757754048 Chr.16: 3728422-3728423 on Build GRCh38
rs745368604 Chr.16: 3728402-3728403 on Build GRCh38
rs992349519 Chr.16: 3728332-3728332 on Build GRCh38
rs763108828 Chr.16: 3728411-3728412 on Build GRCh38
rs769613879 Chr.16: 3728439-3728447 on Build GRCh38
rs750753767 Chr.16: 3728458-3728458 on Build GRCh38
rs775802142 Chr.16: 3728420-3728420 on Build GRCh38
rs772020678 Chr.16: 3728414-3728414 on Build GRCh38
rs770961609 Chr.16: 3728396-3728396 on Build GRCh38
rs146514877 Chr.16: 3728322-3728322 on Build GRCh38
rs760988505 Chr.16: 3728422-3728422 on Build GRCh38
rs3751845 Chr.16: 3728336-3728336 on Build GRCh38
rs764254639 Chr.16: 3728360-3728360 on Build GRCh38
rs747704154 Chr.16: 3728321-3728321 on Build GRCh38
rs55960450 Chr.16: 3728426-3728426 on Build GRCh38
rs142545779 Chr.16: 3728423-3728423 on Build GRCh38
rs746596528 Chr.16: 3728314-3728314 on Build GRCh38
rs769325049 Chr.16: 3728324-3728324 on Build GRCh38
rs776419949 Chr.16: 3728412-3728423 on Build GRCh38
rs759500116 Chr.16: 3728414-3728415 on Build GRCh38
rs764485048 Chr.16: 3728421-3728423 on Build GRCh38
rs768299710 Chr.16: 3728408-3728409 on Build GRCh38
rs750150018 Chr.16: 3728423-3728424 on Build GRCh38
rs759845532 Chr.16: 3728270-3728270 on Build GRCh38
rs750845399 Chr.16: 3728369-3728369 on Build GRCh38
rs777343408 Chr.16: 3728390-3728390 on Build GRCh38
rs774272248 Chr.16: 3728340-3728340 on Build GRCh38
rs1030728642 Chr.16: 3728379-3728379 on Build GRCh38
rs757512971 Chr.16: 3728285-3728285 on Build GRCh38
rs752281964 Chr.16: 3728468-3728468 on Build GRCh38
rs754396803 Chr.16: 3728284-3728284 on Build GRCh38
rs727503890 Chr.16: 3728418-3728426 on Build GRCh38
rs756972103 Chr.16: 3728400-3728402 on Build GRCh38
rs548345138 Chr.16: 3728404-3728404 on Build GRCh38
rs774379679 Chr.16: 3728266-3728266 on Build GRCh38
rs143583342 Chr.16: 3728273-3728273 on Build GRCh38
rs746858489 Chr.16: 3728436-3728438 on Build GRCh38
rs754601626 Chr.16: 3728370-3728370 on Build GRCh38
rs749263945 Chr.16: 3728335-3728335 on Build GRCh38
rs772974749 Chr.16: 3728327-3728327 on Build GRCh38
rs777137937 Chr.16: 3728480-3728480 on Build GRCh38
rs139688311 Chr.16: 3728362-3728362 on Build GRCh38
rs374826846 Chr.16: 3728378-3728378 on Build GRCh38
rs201356785 Chr.16: 3728315-3728315 on Build GRCh38
rs773030588 Chr.16: 3728258-3728258 on Build GRCh38
rs746989288 Chr.16: 3728406-3728414 on Build GRCh38
rs756408732 Chr.16: 3728302-3728304 on Build GRCh38
rs755864452 Chr.16: 3728471-3728471 on Build GRCh38
rs774618955 Chr.16: 3728412-3728414 on Build GRCh38
rs767681669 Chr.16: 3728415-3728429 on Build GRCh38
rs767904307 Chr.16: 3728350-3728350 on Build GRCh38
rs150461438 Chr.16: 3728391-3728391 on Build GRCh38
rs759823393 Chr.16: 3728347-3728347 on Build GRCh38
rs765513337 Chr.16: 3728366-3728366 on Build GRCh38
rs753283086 Chr.16: 3728301-3728302 on Build GRCh38
rs753092630 Chr.16: 3728417-3728418 on Build GRCh38
rs779345611 Chr.16: 3728302-3728302 on Build GRCh38
rs778833800 Chr.16: 3728398-3728398 on Build GRCh38
rs62636220 Chr.16: 3728438-3728438 on Build GRCh38
rs748072121 Chr.16: 3728252-3728252 on Build GRCh38
rs779647460 Chr.16: 3728426-3728427 on Build GRCh38
rs201956327 Chr.16: 3728253-3728253 on Build GRCh38
rs747935597 Chr.16: 3728439-3728444 on Build GRCh38
rs781168285 Chr.16: 3728438-3728439 on Build GRCh38
rs1029488452 Chr.16: 3728289-3728289 on Build GRCh38
rs538605698 Chr.16: 3728448-3728448 on Build GRCh38
rs754267405 Chr.16: 3728421-3728426 on Build GRCh38
rs766744145 Chr.16: 3728467-3728467 on Build GRCh38
rs794727551 Chr.16: 3728301-3728301 on Build GRCh38
rs957150590 Chr.16: 3728328-3728328 on Build GRCh38
rs73491896 Chr.16: 3728483-3728483 on Build GRCh38
rs374379164 Chr.16: 3728261-3728261 on Build GRCh38

View Full Product Details