Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126345150-126345423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCPS
Entrez Gene ID
28960
Gene Name
decapping enzyme, scavenger
Gene Aliases
ARS, DCS1, HINT-5, HINT5, HSL1, HSPC015
UniGene
Hs.504249
Gene Chromosome Location
Chr.11: 126303301-126345754 on Build GRCh38
Transcripts
NM_014026.4 XM_011542778.2 XM_011542780.2 XM_017017612.1 NR_033839.1

Gene Symbol
ST3GAL4-AS1
Entrez Gene ID
399972
Gene Name
ST3GAL4 antisense RNA 1 (head to head)
Gene Aliases
DCPS-AS1
UniGene
Hs.585206
Gene Chromosome Location
Chr.11: 126341716-126355587 on Build GRCh38
Transcripts
NM_014026.4 XM_011542778.2 XM_011542780.2 XM_017017612.1 NR_033839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640984 c.773T>C p.M258T Chr.11: 126345372-126345372 on Build GRCh38
4676579 c.764G>A p.R255H Chr.11: 126345363-126345363 on Build GRCh38
1976135 c.823C>G p.H275D Chr.11: 126345422-126345422 on Build GRCh38
5954557 c.748-6C>T p.? Chr.11: 126345341-126345341 on Build GRCh38
5371978 c.800T>C p.L267P Chr.11: 126345399-126345399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370519459 Chr.11: 126345312-126345312 on Build GRCh38
rs765981711 Chr.11: 126345339-126345339 on Build GRCh38
rs893155245 Chr.11: 126345315-126345315 on Build GRCh38
rs747748685 Chr.11: 126345350-126345350 on Build GRCh38
rs201486424 Chr.11: 126345378-126345378 on Build GRCh38
rs371492107 Chr.11: 126345295-126345295 on Build GRCh38
rs370654094 Chr.11: 126345386-126345386 on Build GRCh38
rs373758046 Chr.11: 126345298-126345298 on Build GRCh38
rs972909120 Chr.11: 126345273-126345273 on Build GRCh38
rs200601649 Chr.11: 126345367-126345367 on Build GRCh38
rs765924535 Chr.11: 126345345-126345347 on Build GRCh38
rs373185234 Chr.11: 126345333-126345333 on Build GRCh38
rs376908736 Chr.11: 126345296-126345296 on Build GRCh38
rs140377449 Chr.11: 126345390-126345390 on Build GRCh38
rs762071568 Chr.11: 126345384-126345384 on Build GRCh38
rs775423831 Chr.11: 126345303-126345303 on Build GRCh38
rs182752869 Chr.11: 126345184-126345184 on Build GRCh38
rs765640045 Chr.11: 126345385-126345385 on Build GRCh38
rs76667195 Chr.11: 126345276-126345276 on Build GRCh38
rs750956205 Chr.11: 126345317-126345317 on Build GRCh38
rs909557110 Chr.11: 126345196-126345196 on Build GRCh38
rs752020530 Chr.11: 126345399-126345399 on Build GRCh38
rs564341931 Chr.11: 126345287-126345287 on Build GRCh38
rs747057647 Chr.11: 126345369-126345369 on Build GRCh38
rs912792065 Chr.11: 126345292-126345292 on Build GRCh38
rs761031435 Chr.11: 126345304-126345304 on Build GRCh38
rs944336022 Chr.11: 126345293-126345293 on Build GRCh38
rs751046484 Chr.11: 126345343-126345343 on Build GRCh38
rs547671658 Chr.11: 126345252-126345252 on Build GRCh38
rs757018526 Chr.11: 126345363-126345363 on Build GRCh38
rs764393255 Chr.11: 126345311-126345311 on Build GRCh38
rs377602692 Chr.11: 126345362-126345362 on Build GRCh38
rs765472444 Chr.11: 126345313-126345313 on Build GRCh38
rs142492066 Chr.11: 126345403-126345403 on Build GRCh38
rs754664727 Chr.11: 126345347-126345347 on Build GRCh38
rs1034279906 Chr.11: 126345393-126345393 on Build GRCh38
rs749310940 Chr.11: 126345366-126345366 on Build GRCh38
rs146794173 Chr.11: 126345368-126345368 on Build GRCh38
rs763573965 Chr.11: 126345389-126345389 on Build GRCh38
rs762236496 Chr.11: 126345335-126345336 on Build GRCh38
rs768819458 Chr.11: 126345371-126345371 on Build GRCh38

View Full Product Details