Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137051204-137051443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD2
Entrez Gene ID
954
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Gene Aliases
CD39L1, NTPDase-2
UniGene
Hs.123036
Gene Chromosome Location
Chr.9: 137048098-137054051 on Build GRCh38
Transcripts
NM_001246.3 NM_203468.2 XM_011519212.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3848101 c.400G>C p.E134Q Chr.9: 137051357-137051357 on Build GRCh38
1461307 c.444C>T p.T148T Chr.9: 137051313-137051313 on Build GRCh38
3926477 c.444C>A p.T148T Chr.9: 137051313-137051313 on Build GRCh38
1685653 c.403delG p.A135fs*9 Chr.9: 137051354-137051354 on Build GRCh38
6654289 c.428T>C p.V143A Chr.9: 137051329-137051329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375537512 Chr.9: 137051413-137051413 on Build GRCh38
rs772794241 Chr.9: 137051264-137051264 on Build GRCh38
rs139027787 Chr.9: 137051284-137051284 on Build GRCh38
rs543401129 Chr.9: 137051422-137051422 on Build GRCh38
rs781198847 Chr.9: 137051229-137051229 on Build GRCh38
rs760409236 Chr.9: 137051270-137051270 on Build GRCh38
rs373230324 Chr.9: 137051362-137051362 on Build GRCh38
rs772644863 Chr.9: 137051348-137051348 on Build GRCh38
rs866913623 Chr.9: 137051371-137051371 on Build GRCh38
rs142095240 Chr.9: 137051316-137051316 on Build GRCh38
rs952543319 Chr.9: 137051307-137051307 on Build GRCh38
rs764953760 Chr.9: 137051373-137051373 on Build GRCh38
rs754031873 Chr.9: 137051412-137051412 on Build GRCh38
rs767588454 Chr.9: 137051290-137051290 on Build GRCh38
rs997771365 Chr.9: 137051320-137051320 on Build GRCh38
rs143374800 Chr.9: 137051277-137051277 on Build GRCh38
rs762486822 Chr.9: 137051233-137051234 on Build GRCh38
rs761717319 Chr.9: 137051324-137051325 on Build GRCh38
rs770757778 Chr.9: 137051353-137051353 on Build GRCh38
rs779154364 Chr.9: 137051418-137051418 on Build GRCh38
rs555069562 Chr.9: 137051259-137051259 on Build GRCh38
rs992897093 Chr.9: 137051363-137051363 on Build GRCh38
rs145805158 Chr.9: 137051324-137051324 on Build GRCh38
rs760364367 Chr.9: 137051378-137051379 on Build GRCh38
rs749636417 Chr.9: 137051242-137051242 on Build GRCh38
rs769936130 Chr.9: 137051234-137051234 on Build GRCh38
rs748652176 Chr.9: 137051258-137051258 on Build GRCh38
rs76730730 Chr.9: 137051225-137051225 on Build GRCh38
rs774142681 Chr.9: 137051246-137051247 on Build GRCh38
rs370985218 Chr.9: 137051383-137051383 on Build GRCh38
rs766408621 Chr.9: 137051411-137051411 on Build GRCh38
rs201198065 Chr.9: 137051244-137051244 on Build GRCh38
rs761509448 Chr.9: 137051314-137051314 on Build GRCh38
rs756382655 Chr.9: 137051294-137051294 on Build GRCh38
rs139476891 Chr.9: 137051379-137051379 on Build GRCh38
rs750844901 Chr.9: 137051334-137051334 on Build GRCh38
rs756152005 Chr.9: 137051408-137051408 on Build GRCh38
rs780458762 Chr.9: 137051238-137051238 on Build GRCh38
rs766798497 Chr.9: 137051349-137051349 on Build GRCh38
rs577721988 Chr.9: 137051393-137051393 on Build GRCh38
rs150531792 Chr.9: 137051370-137051370 on Build GRCh38
rs752597686 Chr.9: 137051285-137051285 on Build GRCh38
rs768216778 Chr.9: 137051275-137051275 on Build GRCh38
rs774691572 Chr.9: 137051252-137051252 on Build GRCh38
rs768782675 Chr.9: 137051238-137051245 on Build GRCh38
rs764001831 Chr.9: 137051398-137051398 on Build GRCh38
rs766126391 Chr.9: 137051272-137051272 on Build GRCh38
rs200918154 Chr.9: 137051293-137051293 on Build GRCh38
rs750547675 Chr.9: 137051404-137051404 on Build GRCh38
rs142498028 Chr.9: 137051345-137051345 on Build GRCh38
rs746103337 Chr.9: 137051233-137051233 on Build GRCh38
rs746505668 Chr.9: 137051350-137051350 on Build GRCh38
rs780223961 Chr.9: 137051306-137051306 on Build GRCh38

View Full Product Details