Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15696373-15696626 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F12
Entrez Gene ID
66002
Gene Name
cytochrome P450 family 4 subfamily F member 12
Gene Aliases
CYPIVF12, F22329_1
UniGene
Hs.131459
Gene Chromosome Location
Chr.19: 15673018-15698819 on Build GRCh38
Transcripts
NM_023944.3 NR_117085.1 XM_006722850.3 XM_011528202.2 XM_011528203.2 XM_011528204.2 XM_011528205.2 XM_011528207.2 XM_011528208.2 XM_017027172.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3529826 c.1385C>T p.S462F Chr.19: 15696500-15696500 on Build GRCh38
5854885 c.1358G>A p.R453K Chr.19: 15696473-15696473 on Build GRCh38
6639371 c.1365T>G p.P455P Chr.19: 15696480-15696480 on Build GRCh38
1197749 c.1336G>T p.D446Y Chr.19: 15696451-15696451 on Build GRCh38
3529824 c.1366C>T p.L456L Chr.19: 15696481-15696481 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748947664 Chr.19: 15696410-15696410 on Build GRCh38
rs757624376 Chr.19: 15696457-15696457 on Build GRCh38
rs868403845 Chr.19: 15696582-15696582 on Build GRCh38
rs764134471 Chr.19: 15696535-15696535 on Build GRCh38
rs746232800 Chr.19: 15696470-15696470 on Build GRCh38
rs752072459 Chr.19: 15696450-15696450 on Build GRCh38
rs755667303 Chr.19: 15696395-15696396 on Build GRCh38
rs200143561 Chr.19: 15696502-15696502 on Build GRCh38
rs1041341843 Chr.19: 15696581-15696581 on Build GRCh38
rs773075246 Chr.19: 15696433-15696433 on Build GRCh38
rs749003945 Chr.19: 15696410-15696412 on Build GRCh38
rs569455285 Chr.19: 15696459-15696459 on Build GRCh38
rs771951701 Chr.19: 15696430-15696430 on Build GRCh38
rs199619253 Chr.19: 15696425-15696425 on Build GRCh38
rs201640448 Chr.19: 15696520-15696520 on Build GRCh38
rs1034183700 Chr.19: 15696603-15696603 on Build GRCh38
rs751672892 Chr.19: 15696551-15696551 on Build GRCh38
rs1023281461 Chr.19: 15696557-15696557 on Build GRCh38
rs139253757 Chr.19: 15696588-15696588 on Build GRCh38
rs771239208 Chr.19: 15696403-15696403 on Build GRCh38
rs747637199 Chr.19: 15696485-15696485 on Build GRCh38
rs778560842 Chr.19: 15696476-15696476 on Build GRCh38
rs529430043 Chr.19: 15696439-15696439 on Build GRCh38
rs201554483 Chr.19: 15696429-15696429 on Build GRCh38
rs765562898 Chr.19: 15696440-15696440 on Build GRCh38
rs138003903 Chr.19: 15696419-15696419 on Build GRCh38
rs747926643 Chr.19: 15696563-15696564 on Build GRCh38
rs774182052 Chr.19: 15696525-15696526 on Build GRCh38
rs371356525 Chr.19: 15696513-15696513 on Build GRCh38
rs368954465 Chr.19: 15696443-15696443 on Build GRCh38
rs571996132 Chr.19: 15696517-15696517 on Build GRCh38
rs147287420 Chr.19: 15696587-15696587 on Build GRCh38
rs377249673 Chr.19: 15696438-15696438 on Build GRCh38
rs779385565 Chr.19: 15696406-15696406 on Build GRCh38
rs200582035 Chr.19: 15696435-15696435 on Build GRCh38
rs893685583 Chr.19: 15696428-15696428 on Build GRCh38
rs746981355 Chr.19: 15696501-15696501 on Build GRCh38
rs17853419 Chr.19: 15696497-15696497 on Build GRCh38
rs596252 Chr.19: 15696402-15696402 on Build GRCh38
rs757638040 Chr.19: 15696552-15696552 on Build GRCh38
rs969154348 Chr.19: 15696575-15696575 on Build GRCh38
rs200290961 Chr.19: 15696397-15696397 on Build GRCh38
rs996200678 Chr.19: 15696496-15696496 on Build GRCh38
rs112002618 Chr.19: 15696521-15696521 on Build GRCh38
rs973515786 Chr.19: 15696548-15696548 on Build GRCh38
rs751788413 Chr.19: 15696486-15696486 on Build GRCh38
rs779752001 Chr.19: 15696408-15696408 on Build GRCh38
rs374175073 Chr.19: 15696500-15696500 on Build GRCh38
rs754582892 Chr.19: 15696473-15696473 on Build GRCh38
rs773781666 Chr.19: 15696423-15696423 on Build GRCh38
rs768098978 Chr.19: 15696556-15696556 on Build GRCh38
rs376379053 Chr.19: 15696446-15696446 on Build GRCh38
rs373149297 Chr.19: 15696445-15696445 on Build GRCh38
rs963652687 Chr.19: 15696524-15696524 on Build GRCh38
rs688755 Chr.19: 15696495-15696495 on Build GRCh38
rs768164881 Chr.19: 15696414-15696414 on Build GRCh38
rs767492960 Chr.19: 15696401-15696401 on Build GRCh38

View Full Product Details