Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150056048-150056295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF1R
Entrez Gene ID
1436
Gene Name
colony stimulating factor 1 receptor
Gene Aliases
C-FMS, CD115, CSF-1R, CSFR, FIM2, FMS, HDLS, M-CSF-R
UniGene
Hs.586219
Gene Chromosome Location
Chr.5: 150053291-150113372 on Build GRCh38
Transcripts
NM_001288705.1 NM_005211.3 NR_109969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6102842 c.2429T>C p.I810T Chr.5: 150056232-150056232 on Build GRCh38
1435123 c.2512G>A p.V838I Chr.5: 150056068-150056068 on Build GRCh38
1696550 c.2366G>A p.G789D Chr.5: 150056295-150056295 on Build GRCh38
3612423 c.2509G>A p.D837N Chr.5: 150056071-150056071 on Build GRCh38
6968553 c.2442T>A p.N814K Chr.5: 150056219-150056219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs690016557 Chr.5: 150056068-150056068 on Build GRCh38
rs690016550 Chr.5: 150056100-150056100 on Build GRCh38
rs774843893 Chr.5: 150056156-150056156 on Build GRCh38
rs890792389 Chr.5: 150056147-150056147 on Build GRCh38
rs754928706 Chr.5: 150056188-150056188 on Build GRCh38
rs777479233 Chr.5: 150056228-150056228 on Build GRCh38
rs375438101 Chr.5: 150056164-150056164 on Build GRCh38
rs281860275 Chr.5: 150056214-150056214 on Build GRCh38
rs758314616 Chr.5: 150056069-150056069 on Build GRCh38
rs216136 Chr.5: 150056196-150056196 on Build GRCh38
rs771408942 Chr.5: 150056143-150056143 on Build GRCh38
rs1007084965 Chr.5: 150056149-150056149 on Build GRCh38
rs755763539 Chr.5: 150056105-150056105 on Build GRCh38
rs690016549 Chr.5: 150056130-150056130 on Build GRCh38
rs751573716 Chr.5: 150056183-150056183 on Build GRCh38
rs776564056 Chr.5: 150056154-150056155 on Build GRCh38
rs767179933 Chr.5: 150056191-150056191 on Build GRCh38
rs747837811 Chr.5: 150056096-150056096 on Build GRCh38
rs367984340 Chr.5: 150056225-150056225 on Build GRCh38
rs751178536 Chr.5: 150056080-150056080 on Build GRCh38
rs764353006 Chr.5: 150056170-150056170 on Build GRCh38
rs531754207 Chr.5: 150056120-150056120 on Build GRCh38
rs772300724 Chr.5: 150056166-150056166 on Build GRCh38
rs1018009623 Chr.5: 150056160-150056160 on Build GRCh38
rs397515557 Chr.5: 150056097-150056097 on Build GRCh38
rs759339969 Chr.5: 150056173-150056174 on Build GRCh38
rs368711941 Chr.5: 150056167-150056167 on Build GRCh38
rs760846210 Chr.5: 150056169-150056169 on Build GRCh38
rs766207127 Chr.5: 150056180-150056180 on Build GRCh38
rs690016562 Chr.5: 150056112-150056112 on Build GRCh38
rs780804532 Chr.5: 150056082-150056082 on Build GRCh38
rs748825247 Chr.5: 150056245-150056245 on Build GRCh38
rs149872699 Chr.5: 150056081-150056081 on Build GRCh38
rs387906662 Chr.5: 150056071-150056071 on Build GRCh38
rs6880954 Chr.5: 150056171-150056171 on Build GRCh38
rs778050359 Chr.5: 150056108-150056108 on Build GRCh38
rs587777246 Chr.5: 150056218-150056218 on Build GRCh38
rs775508236 Chr.5: 150056168-150056168 on Build GRCh38
rs765139248 Chr.5: 150056176-150056177 on Build GRCh38
rs146691087 Chr.5: 150056072-150056072 on Build GRCh38

View Full Product Details