Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49018076-49018317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DALRD3
Entrez Gene ID
55152
Gene Name
DALR anticodon binding domain containing 3
Gene Aliases
-
UniGene
Hs.654815
Gene Chromosome Location
Chr.3: 49015399-49021386 on Build GRCh38
Transcripts
NM_001009996.2 NM_001276405.1 NM_018114.5 XM_005265269.3 XM_006713219.2 XM_011533891.1 XM_011533892.1 XM_011533893.1 XM_017006723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3595167 c.295C>T p.L99F Chr.3: 49018189-49018189 on Build GRCh38
6421497 c.289C>T p.R97C Chr.3: 49018195-49018195 on Build GRCh38
6385462 c.233G>T p.C78F Chr.3: 49018251-49018251 on Build GRCh38
6038611 c.202C>T p.Q68* Chr.3: 49018282-49018282 on Build GRCh38
6362250 c.322C>T p.P108S Chr.3: 49018162-49018162 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773902952 Chr.3: 49018106-49018106 on Build GRCh38
rs948896947 Chr.3: 49018198-49018198 on Build GRCh38
rs767582011 Chr.3: 49018130-49018130 on Build GRCh38
rs770338797 Chr.3: 49018244-49018244 on Build GRCh38
rs748420382 Chr.3: 49018098-49018098 on Build GRCh38
rs973848801 Chr.3: 49018152-49018152 on Build GRCh38
rs918522039 Chr.3: 49018163-49018163 on Build GRCh38
rs753833477 Chr.3: 49018157-49018157 on Build GRCh38
rs777850206 Chr.3: 49018228-49018229 on Build GRCh38
rs771557967 Chr.3: 49018093-49018093 on Build GRCh38
rs1052746715 Chr.3: 49018265-49018265 on Build GRCh38
rs759813973 Chr.3: 49018126-49018126 on Build GRCh38
rs868425035 Chr.3: 49018116-49018116 on Build GRCh38
rs778399350 Chr.3: 49018175-49018175 on Build GRCh38
rs560044928 Chr.3: 49018219-49018219 on Build GRCh38
rs749814901 Chr.3: 49018295-49018295 on Build GRCh38
rs749734472 Chr.3: 49018259-49018269 on Build GRCh38
rs752571745 Chr.3: 49018132-49018132 on Build GRCh38
rs1012175222 Chr.3: 49018246-49018246 on Build GRCh38
rs749846952 Chr.3: 49018180-49018180 on Build GRCh38
rs1056310048 Chr.3: 49018225-49018225 on Build GRCh38
rs757183504 Chr.3: 49018174-49018174 on Build GRCh38
rs1041866651 Chr.3: 49018258-49018258 on Build GRCh38
rs774226234 Chr.3: 49018123-49018123 on Build GRCh38
rs758100563 Chr.3: 49018196-49018196 on Build GRCh38
rs778275864 Chr.3: 49018264-49018264 on Build GRCh38
rs563509705 Chr.3: 49018160-49018160 on Build GRCh38
rs972011922 Chr.3: 49018146-49018146 on Build GRCh38
rs781630790 Chr.3: 49018097-49018097 on Build GRCh38
rs763856713 Chr.3: 49018150-49018150 on Build GRCh38
rs746649290 Chr.3: 49018223-49018223 on Build GRCh38
rs892786633 Chr.3: 49018272-49018272 on Build GRCh38
rs1028648261 Chr.3: 49018284-49018284 on Build GRCh38
rs375078266 Chr.3: 49018154-49018154 on Build GRCh38
rs937339146 Chr.3: 49018208-49018208 on Build GRCh38
rs763538951 Chr.3: 49018115-49018115 on Build GRCh38
rs11542202 Chr.3: 49018239-49018239 on Build GRCh38
rs1011518563 Chr.3: 49018276-49018276 on Build GRCh38
rs930934806 Chr.3: 49018168-49018168 on Build GRCh38
rs746296701 Chr.3: 49018134-49018134 on Build GRCh38
rs906916343 Chr.3: 49018234-49018234 on Build GRCh38
rs928665276 Chr.3: 49018185-49018185 on Build GRCh38
rs941220460 Chr.3: 49018140-49018140 on Build GRCh38
rs771199097 Chr.3: 49018118-49018118 on Build GRCh38
rs916115883 Chr.3: 49018195-49018195 on Build GRCh38

View Full Product Details