Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 28494795-28495068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB5
Entrez Gene ID
9586
Gene Name
cAMP responsive element binding protein 5
Gene Aliases
CRE-BPA, CREB-5
UniGene
Hs.437075
Gene Chromosome Location
Chr.7: 28299321-28825894 on Build GRCh38
Transcripts
NM_004904.3 NM_182898.3 NM_182899.4 XM_005249906.1 XM_017012806.1 XM_017012807.1 XM_017012808.1 XM_017012809.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3950383 c.76-6C>G p.? Chr.7: 28494900-28494900 on Build GRCh38
3637780 c.121G>A p.E41K Chr.7: 28494951-28494951 on Build GRCh38
6635332 c.85A>G p.T29A Chr.7: 28494915-28494915 on Build GRCh38
3637779 c.81C>T p.F27F Chr.7: 28494911-28494911 on Build GRCh38
5599341 c.140C>T p.P47L Chr.7: 28494970-28494970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769485767 Chr.7: 28494961-28494961 on Build GRCh38
rs770744190 Chr.7: 28494874-28494874 on Build GRCh38
rs540044957 Chr.7: 28494879-28494879 on Build GRCh38
rs754986749 Chr.7: 28494906-28494906 on Build GRCh38
rs377532708 Chr.7: 28494872-28494872 on Build GRCh38
rs1055784864 Chr.7: 28494965-28494965 on Build GRCh38
rs1012091745 Chr.7: 28494824-28494824 on Build GRCh38
rs759069269 Chr.7: 28494884-28494884 on Build GRCh38
rs3216916 Chr.7: 28494883-28494884 on Build GRCh38
rs769572302 Chr.7: 28494893-28494893 on Build GRCh38
rs893266857 Chr.7: 28494823-28494823 on Build GRCh38
rs762985710 Chr.7: 28494924-28494926 on Build GRCh38
rs760887381 Chr.7: 28495015-28495015 on Build GRCh38
rs957771900 Chr.7: 28494848-28494848 on Build GRCh38
rs869183954 Chr.7: 28494893-28494894 on Build GRCh38
rs775348413 Chr.7: 28494896-28494896 on Build GRCh38
rs781219951 Chr.7: 28494944-28494944 on Build GRCh38
rs757241986 Chr.7: 28494933-28494933 on Build GRCh38
rs77306029 Chr.7: 28494900-28494900 on Build GRCh38
rs776313709 Chr.7: 28494880-28494880 on Build GRCh38
rs766879037 Chr.7: 28495019-28495019 on Build GRCh38
rs777663588 Chr.7: 28494924-28494924 on Build GRCh38
rs777192547 Chr.7: 28495024-28495024 on Build GRCh38
rs752978846 Chr.7: 28494908-28494908 on Build GRCh38
rs539813790 Chr.7: 28494880-28494881 on Build GRCh38
rs764412503 Chr.7: 28494992-28494992 on Build GRCh38
rs141934847 Chr.7: 28494917-28494917 on Build GRCh38
rs80296494 Chr.7: 28494973-28494973 on Build GRCh38
rs749010019 Chr.7: 28494986-28494986 on Build GRCh38
rs771142919 Chr.7: 28494884-28494884 on Build GRCh38
rs373496439 Chr.7: 28494897-28494897 on Build GRCh38
rs777420837 Chr.7: 28494868-28494868 on Build GRCh38
rs758682194 Chr.7: 28495032-28495032 on Build GRCh38
rs369761252 Chr.7: 28494871-28494871 on Build GRCh38
rs528332433 Chr.7: 28494866-28494866 on Build GRCh38
rs747943829 Chr.7: 28494864-28494864 on Build GRCh38
rs765350331 Chr.7: 28495028-28495028 on Build GRCh38
rs397696722 Chr.7: 28494892-28494893 on Build GRCh38
rs746742237 Chr.7: 28494930-28494930 on Build GRCh38
rs370491584 Chr.7: 28495027-28495027 on Build GRCh38
rs752747806 Chr.7: 28495030-28495030 on Build GRCh38
rs1020338588 Chr.7: 28494846-28494846 on Build GRCh38
rs775117095 Chr.7: 28494975-28494975 on Build GRCh38
rs765299339 Chr.7: 28494907-28494907 on Build GRCh38
rs963222016 Chr.7: 28494920-28494920 on Build GRCh38
rs763712573 Chr.7: 28494921-28494921 on Build GRCh38
rs752821429 Chr.7: 28494896-28494897 on Build GRCh38
rs530130087 Chr.7: 28494987-28494987 on Build GRCh38
rs373211496 Chr.7: 28494903-28494903 on Build GRCh38
rs766744683 Chr.7: 28494901-28494901 on Build GRCh38
rs200825206 Chr.7: 28494899-28494899 on Build GRCh38
rs569232806 Chr.7: 28494937-28494937 on Build GRCh38
rs376480957 Chr.7: 28495011-28495011 on Build GRCh38
rs745795118 Chr.7: 28494956-28494956 on Build GRCh38

View Full Product Details