Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96304281-96304542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNTT
Entrez Gene ID
1791
Gene Name
DNA nucleotidylexotransferase
Gene Aliases
TDT
UniGene
Hs.534206
Gene Chromosome Location
Chr.10: 96304328-96338564 on Build GRCh38
Transcripts
NM_001017520.1 NM_004088.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4546424 c.3G>A p.M1I Chr.10: 96304500-96304500 on Build GRCh38
4807876 c.18G>A p.A6A Chr.10: 96304515-96304515 on Build GRCh38
5627992 c.39G>A p.K13K Chr.10: 96304536-96304536 on Build GRCh38
3808105 c.14G>A p.R5Q Chr.10: 96304511-96304511 on Build GRCh38
304511 c.34C>T p.R12W Chr.10: 96304531-96304531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774258749 Chr.10: 96304515-96304515 on Build GRCh38
rs937969443 Chr.10: 96304395-96304395 on Build GRCh38
rs758918471 Chr.10: 96304457-96304457 on Build GRCh38
rs1001047247 Chr.10: 96304444-96304444 on Build GRCh38
rs557814941 Chr.10: 96304419-96304419 on Build GRCh38
rs144113836 Chr.10: 96304507-96304507 on Build GRCh38
rs781536667 Chr.10: 96304498-96304498 on Build GRCh38
rs771027503 Chr.10: 96304514-96304514 on Build GRCh38
rs769845607 Chr.10: 96304506-96304506 on Build GRCh38
rs572802954 Chr.10: 96304445-96304445 on Build GRCh38
rs538354192 Chr.10: 96304330-96304330 on Build GRCh38
rs1056474188 Chr.10: 96304397-96304397 on Build GRCh38
rs962189183 Chr.10: 96304443-96304443 on Build GRCh38
rs1014212035 Chr.10: 96304390-96304390 on Build GRCh38
rs1021240633 Chr.10: 96304422-96304422 on Build GRCh38
rs748174871 Chr.10: 96304502-96304502 on Build GRCh38
rs1034244037 Chr.10: 96304325-96304325 on Build GRCh38
rs747403482 Chr.10: 96304469-96304469 on Build GRCh38
rs1024226734 Chr.10: 96304414-96304414 on Build GRCh38
rs755122304 Chr.10: 96304479-96304479 on Build GRCh38
rs780347987 Chr.10: 96304468-96304468 on Build GRCh38
rs757569416 Chr.10: 96304510-96304510 on Build GRCh38
rs952300548 Chr.10: 96304511-96304511 on Build GRCh38
rs958593651 Chr.10: 96304326-96304326 on Build GRCh38
rs1038387220 Chr.10: 96304361-96304361 on Build GRCh38

View Full Product Details