Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 52657997-52658248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERO1A
Entrez Gene ID
30001
Gene Name
endoplasmic reticulum oxidoreductase 1 alpha
Gene Aliases
ERO1-L, ERO1-L-alpha, ERO1-alpha, ERO1L, ERO1LA, Ero1alpha
UniGene
Hs.525339
Gene Chromosome Location
Chr.14: 52639915-52695931 on Build GRCh38
Transcripts
NM_014584.2 XM_011536684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6463564 c.689-7_689-4delATTT p.? Chr.14: 52658154-52658157 on Build GRCh38
260724 c.718C>A p.L240I Chr.14: 52658007-52658007 on Build GRCh38
6458830 c.689-2_689-1insGGAG p.? Chr.14: 52658151-52658152 on Build GRCh38
2199598 c.716G>T p.G239V Chr.14: 52658009-52658009 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs62639686 Chr.14: 52658149-52658149 on Build GRCh38
rs376003500 Chr.14: 52658117-52658117 on Build GRCh38
rs766137746 Chr.14: 52658081-52658081 on Build GRCh38
rs751208796 Chr.14: 52658157-52658157 on Build GRCh38
rs775352345 Chr.14: 52658185-52658185 on Build GRCh38
rs767578735 Chr.14: 52658185-52658193 on Build GRCh38
rs377241560 Chr.14: 52658088-52658089 on Build GRCh38
rs1051427608 Chr.14: 52658109-52658109 on Build GRCh38
rs995471510 Chr.14: 52658221-52658221 on Build GRCh38
rs776899795 Chr.14: 52658051-52658051 on Build GRCh38
rs773447036 Chr.14: 52658101-52658105 on Build GRCh38
rs757831532 Chr.14: 52658143-52658143 on Build GRCh38
rs768309206 Chr.14: 52658044-52658044 on Build GRCh38
rs74832745 Chr.14: 52658055-52658055 on Build GRCh38
rs963650938 Chr.14: 52658176-52658176 on Build GRCh38
rs535398223 Chr.14: 52658199-52658199 on Build GRCh38
rs754651346 Chr.14: 52658161-52658161 on Build GRCh38
rs762659542 Chr.14: 52658077-52658077 on Build GRCh38
rs773124743 Chr.14: 52658071-52658071 on Build GRCh38
rs761618726 Chr.14: 52658077-52658078 on Build GRCh38
rs564257920 Chr.14: 52658060-52658060 on Build GRCh38
rs549898349 Chr.14: 52658095-52658095 on Build GRCh38
rs753901014 Chr.14: 52658138-52658138 on Build GRCh38
rs769363035 Chr.14: 52658202-52658202 on Build GRCh38
rs769953480 Chr.14: 52658081-52658082 on Build GRCh38
rs746626746 Chr.14: 52658023-52658023 on Build GRCh38
rs750887051 Chr.14: 52658147-52658147 on Build GRCh38
rs768970050 Chr.14: 52658044-52658045 on Build GRCh38
rs748473341 Chr.14: 52658210-52658211 on Build GRCh38
rs776248167 Chr.14: 52658047-52658047 on Build GRCh38
rs890238334 Chr.14: 52658125-52658125 on Build GRCh38
rs765743043 Chr.14: 52658145-52658145 on Build GRCh38
rs747656385 Chr.14: 52658201-52658201 on Build GRCh38
rs3783453 Chr.14: 52658089-52658089 on Build GRCh38
rs959555429 Chr.14: 52658093-52658093 on Build GRCh38
rs763172885 Chr.14: 52658170-52658171 on Build GRCh38

View Full Product Details