Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 41346956-41347189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3B
Entrez Gene ID
54097
Gene Name
family with sequence similarity 3 member B
Gene Aliases
2-21, C21orf11, C21orf76, ORF9, PANDER, PRED44
UniGene
Hs.473877
Gene Chromosome Location
Chr.21: 41304229-41357727 on Build GRCh38
Transcripts
NM_058186.3 NM_206964.1 XM_011529648.2 XM_011529649.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030847 c.434C>T p.A145V Chr.21: 41347049-41347049 on Build GRCh38
4101677 c.432T>C p.S144S Chr.21: 41347047-41347047 on Build GRCh38
6341282 c.485+8G>T p.? Chr.21: 41347108-41347108 on Build GRCh38
4549564 c.476G>A p.G159E Chr.21: 41347091-41347091 on Build GRCh38
1414175 c.474C>T p.D158D Chr.21: 41347089-41347089 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768234423 Chr.21: 41347090-41347090 on Build GRCh38
rs768309277 Chr.21: 41347001-41347001 on Build GRCh38
rs990516469 Chr.21: 41347145-41347145 on Build GRCh38
rs573794470 Chr.21: 41347089-41347089 on Build GRCh38
rs754095511 Chr.21: 41347142-41347142 on Build GRCh38
rs768017285 Chr.21: 41347123-41347123 on Build GRCh38
rs753872210 Chr.21: 41347048-41347048 on Build GRCh38
rs746658582 Chr.21: 41346983-41346983 on Build GRCh38
rs749193689 Chr.21: 41347108-41347108 on Build GRCh38
rs185024705 Chr.21: 41347151-41347151 on Build GRCh38
rs190096507 Chr.21: 41347126-41347126 on Build GRCh38
rs139452370 Chr.21: 41347025-41347025 on Build GRCh38
rs972647937 Chr.21: 41347169-41347169 on Build GRCh38
rs368256800 Chr.21: 41347121-41347121 on Build GRCh38
rs772641781 Chr.21: 41347118-41347118 on Build GRCh38
rs756310800 Chr.21: 41347035-41347035 on Build GRCh38
rs753047922 Chr.21: 41347028-41347028 on Build GRCh38
rs745807748 Chr.21: 41347074-41347074 on Build GRCh38
rs148783965 Chr.21: 41347084-41347084 on Build GRCh38
rs942382717 Chr.21: 41347082-41347082 on Build GRCh38
rs962640317 Chr.21: 41347168-41347168 on Build GRCh38
rs202154015 Chr.21: 41347009-41347009 on Build GRCh38
rs774827145 Chr.21: 41347024-41347024 on Build GRCh38
rs766325526 Chr.21: 41347016-41347017 on Build GRCh38
rs776326598 Chr.21: 41347098-41347098 on Build GRCh38
rs145464636 Chr.21: 41347073-41347073 on Build GRCh38
rs182292891 Chr.21: 41347127-41347127 on Build GRCh38
rs775292352 Chr.21: 41346982-41346982 on Build GRCh38
rs766922280 Chr.21: 41347010-41347010 on Build GRCh38
rs757385893 Chr.21: 41347057-41347057 on Build GRCh38
rs144944684 Chr.21: 41347086-41347086 on Build GRCh38
rs779095124 Chr.21: 41347087-41347087 on Build GRCh38
rs753027099 Chr.21: 41347017-41347018 on Build GRCh38
rs371998204 Chr.21: 41347109-41347109 on Build GRCh38
rs753952994 Chr.21: 41347116-41347116 on Build GRCh38
rs149676623 Chr.21: 41347026-41347026 on Build GRCh38
rs376984399 Chr.21: 41347116-41347116 on Build GRCh38
rs199567406 Chr.21: 41347140-41347140 on Build GRCh38

View Full Product Details