Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37621505-37621778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGA1
Entrez Gene ID
26088
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 1
Gene Aliases
-
UniGene
Hs.499158
Gene Chromosome Location
Chr.22: 37607235-37633564 on Build GRCh38
Transcripts
NM_001001560.2 NM_001172687.1 NM_001172688.1 NM_013365.4 XM_005261517.3 XM_005261518.1 XM_005261520.1 XM_005261522.1 XM_006724229.1 XM_011530122.1 XM_011530123.1 XM_011530124.1 XM_017028760.1 XM_017028761.1 XM_017028762.1 XM_017028763.1 XM_017028764.1 XM_017028765.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3842636 c.574G>A p.A192T Chr.22: 37621661-37621661 on Build GRCh38
5105262 c.586C>A p.L196I Chr.22: 37621673-37621673 on Build GRCh38
6384975 c.556C>A p.H186N Chr.22: 37621643-37621643 on Build GRCh38
1034022 c.560C>T p.P187L Chr.22: 37621647-37621647 on Build GRCh38
4798773 c.561C>G p.P187P Chr.22: 37621648-37621648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530201995 Chr.22: 37621599-37621599 on Build GRCh38
rs571543713 Chr.22: 37621573-37621573 on Build GRCh38
rs944520533 Chr.22: 37621543-37621543 on Build GRCh38
rs756104307 Chr.22: 37621693-37621693 on Build GRCh38
rs370188432 Chr.22: 37621535-37621535 on Build GRCh38
rs1002304134 Chr.22: 37621658-37621658 on Build GRCh38
rs1033425098 Chr.22: 37621661-37621661 on Build GRCh38
rs375317914 Chr.22: 37621647-37621647 on Build GRCh38
rs772837405 Chr.22: 37621593-37621593 on Build GRCh38
rs761679802 Chr.22: 37621716-37621719 on Build GRCh38
rs747882330 Chr.22: 37621718-37621718 on Build GRCh38
rs372607959 Chr.22: 37621711-37621711 on Build GRCh38
rs373435349 Chr.22: 37621616-37621616 on Build GRCh38
rs750517578 Chr.22: 37621688-37621688 on Build GRCh38
rs937350026 Chr.22: 37621597-37621597 on Build GRCh38
rs900137780 Chr.22: 37621752-37621752 on Build GRCh38
rs757557099 Chr.22: 37621660-37621660 on Build GRCh38
rs569212065 Chr.22: 37621648-37621648 on Build GRCh38
rs768686778 Chr.22: 37621705-37621705 on Build GRCh38
rs369010190 Chr.22: 37621702-37621702 on Build GRCh38
rs960481770 Chr.22: 37621669-37621669 on Build GRCh38
rs765739293 Chr.22: 37621610-37621610 on Build GRCh38
rs189717909 Chr.22: 37621625-37621625 on Build GRCh38
rs1040194052 Chr.22: 37621546-37621546 on Build GRCh38
rs138483740 Chr.22: 37621675-37621675 on Build GRCh38
rs1015679974 Chr.22: 37621708-37621708 on Build GRCh38
rs866195916 Chr.22: 37621551-37621551 on Build GRCh38
rs982139478 Chr.22: 37621575-37621575 on Build GRCh38
rs375670105 Chr.22: 37621591-37621591 on Build GRCh38
rs774272074 Chr.22: 37621566-37621567 on Build GRCh38
rs1040350443 Chr.22: 37621747-37621747 on Build GRCh38
rs201580085 Chr.22: 37621592-37621592 on Build GRCh38
rs200605527 Chr.22: 37621730-37621730 on Build GRCh38
rs760046335 Chr.22: 37621601-37621601 on Build GRCh38
rs751865774 Chr.22: 37621649-37621649 on Build GRCh38
rs771770872 Chr.22: 37621729-37621729 on Build GRCh38
rs760265718 Chr.22: 37621738-37621738 on Build GRCh38
rs370627329 Chr.22: 37621743-37621743 on Build GRCh38

View Full Product Details