Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 29128664-29128937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM189A1
Entrez Gene ID
23359
Gene Name
family with sequence similarity 189 member A1
Gene Aliases
TMEM228
UniGene
Hs.383564
Gene Chromosome Location
Chr.15: 29120252-29570723 on Build GRCh38
Transcripts
NM_015307.1 XM_011521407.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4417919 c.981C>G p.T327T Chr.15: 29128811-29128811 on Build GRCh38
4417852 c.941G>A p.G314D Chr.15: 29128851-29128851 on Build GRCh38
1608226 c.987T>A p.A329A Chr.15: 29128805-29128805 on Build GRCh38
4751857 c.934A>G p.S312G Chr.15: 29128858-29128858 on Build GRCh38
4975307 c.954C>T p.A318A Chr.15: 29128838-29128838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs191724006 Chr.15: 29128902-29128902 on Build GRCh38
rs771963029 Chr.15: 29128893-29128893 on Build GRCh38
rs193244349 Chr.15: 29128694-29128694 on Build GRCh38
rs778670252 Chr.15: 29128861-29128861 on Build GRCh38
rs556050354 Chr.15: 29128687-29128687 on Build GRCh38
rs954812931 Chr.15: 29128803-29128803 on Build GRCh38
rs201612440 Chr.15: 29128825-29128825 on Build GRCh38
rs993770928 Chr.15: 29128756-29128756 on Build GRCh38
rs539511502 Chr.15: 29128696-29128697 on Build GRCh38
rs1018037845 Chr.15: 29128812-29128812 on Build GRCh38
rs34148521 Chr.15: 29128880-29128881 on Build GRCh38
rs766779801 Chr.15: 29128838-29128838 on Build GRCh38
rs112409522 Chr.15: 29128725-29128725 on Build GRCh38
rs993243629 Chr.15: 29128911-29128911 on Build GRCh38
rs386782512 Chr.15: 29128725-29128727 on Build GRCh38
rs978845028 Chr.15: 29128822-29128822 on Build GRCh38
rs75662648 Chr.15: 29128720-29128720 on Build GRCh38
rs374164421 Chr.15: 29128886-29128886 on Build GRCh38
rs117801457 Chr.15: 29128910-29128910 on Build GRCh38
rs200682124 Chr.15: 29128856-29128856 on Build GRCh38
rs776793558 Chr.15: 29128793-29128793 on Build GRCh38
rs765541310 Chr.15: 29128817-29128817 on Build GRCh38
rs775220949 Chr.15: 29128894-29128894 on Build GRCh38
rs764161882 Chr.15: 29128765-29128765 on Build GRCh38
rs545288833 Chr.15: 29128704-29128704 on Build GRCh38
rs1014489335 Chr.15: 29128691-29128691 on Build GRCh38
rs752079448 Chr.15: 29128731-29128731 on Build GRCh38
rs755619498 Chr.15: 29128849-29128849 on Build GRCh38
rs143753846 Chr.15: 29128692-29128696 on Build GRCh38
rs745311647 Chr.15: 29128896-29128897 on Build GRCh38
rs148130805 Chr.15: 29128727-29128727 on Build GRCh38
rs564368583 Chr.15: 29128719-29128719 on Build GRCh38
rs768670571 Chr.15: 29128907-29128907 on Build GRCh38
rs548097301 Chr.15: 29128691-29128692 on Build GRCh38
rs1053947323 Chr.15: 29128695-29128695 on Build GRCh38
rs1037319089 Chr.15: 29128903-29128903 on Build GRCh38
rs904838250 Chr.15: 29128708-29128708 on Build GRCh38
rs752127270 Chr.15: 29128848-29128848 on Build GRCh38
rs758861932 Chr.15: 29128837-29128837 on Build GRCh38
rs2306934 Chr.15: 29128811-29128811 on Build GRCh38
rs186472907 Chr.15: 29128762-29128762 on Build GRCh38
rs200482015 Chr.15: 29128885-29128885 on Build GRCh38
rs748901080 Chr.15: 29128854-29128854 on Build GRCh38
rs2306933 Chr.15: 29128851-29128851 on Build GRCh38

View Full Product Details