Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48878399-48878638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EID1
Entrez Gene ID
23741
Gene Name
EP300 interacting inhibitor of differentiation 1
Gene Aliases
C15orf3, CRI1, EID-1, IRO45620, PNAS-22, PTD014, RBP21
UniGene
Hs.255973
Gene Chromosome Location
Chr.15: 48878093-48880183 on Build GRCh38
Transcripts
NM_014335.2 NM_203349.3 XM_005254375.3

Gene Symbol
SHC4
Entrez Gene ID
399694
Gene Name
SHC adaptor protein 4
Gene Aliases
RaLP, SHCD
UniGene
Hs.642615
Gene Chromosome Location
Chr.15: 48823737-48963444 on Build GRCh38
Transcripts
NM_014335.2 NM_203349.3 XM_005254375.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1608452 c.446T>A p.M149K Chr.15: 48878622-48878622 on Build GRCh38
5095204 c.239A>T p.N80I Chr.15: 48878415-48878415 on Build GRCh38
2216061 c.454G>A p.E152K Chr.15: 48878630-48878630 on Build GRCh38
4428513 c.413C>G p.T138R Chr.15: 48878589-48878589 on Build GRCh38
1587906 c.300G>T p.E100D Chr.15: 48878476-48878476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764215628 Chr.15: 48878444-48878444 on Build GRCh38
rs527861826 Chr.15: 48878470-48878470 on Build GRCh38
rs538774168 Chr.15: 48878545-48878545 on Build GRCh38
rs766741446 Chr.15: 48878548-48878548 on Build GRCh38
rs779336657 Chr.15: 48878455-48878455 on Build GRCh38
rs865905112 Chr.15: 48878568-48878568 on Build GRCh38
rs76033918 Chr.15: 48878600-48878600 on Build GRCh38
rs1015274737 Chr.15: 48878501-48878501 on Build GRCh38
rs747676847 Chr.15: 48878495-48878495 on Build GRCh38
rs780719516 Chr.15: 48878490-48878490 on Build GRCh38
rs1028481351 Chr.15: 48878436-48878436 on Build GRCh38
rs761926680 Chr.15: 48878534-48878534 on Build GRCh38
rs556390836 Chr.15: 48878513-48878513 on Build GRCh38
rs770532054 Chr.15: 48878597-48878597 on Build GRCh38
rs368986494 Chr.15: 48878530-48878530 on Build GRCh38
rs753869143 Chr.15: 48878585-48878590 on Build GRCh38
rs564257429 Chr.15: 48878473-48878473 on Build GRCh38
rs1021880299 Chr.15: 48878481-48878481 on Build GRCh38
rs948549137 Chr.15: 48878591-48878591 on Build GRCh38
rs755502226 Chr.15: 48878564-48878564 on Build GRCh38
rs751908848 Chr.15: 48878557-48878557 on Build GRCh38
rs750687807 Chr.15: 48878456-48878456 on Build GRCh38
rs12914632 Chr.15: 48878594-48878594 on Build GRCh38
rs770668281 Chr.15: 48878515-48878515 on Build GRCh38
rs112644597 Chr.15: 48878503-48878503 on Build GRCh38
rs774065138 Chr.15: 48878521-48878521 on Build GRCh38
rs770806745 Chr.15: 48878524-48878524 on Build GRCh38
rs566866767 Chr.15: 48878442-48878442 on Build GRCh38
rs757550039 Chr.15: 48878451-48878451 on Build GRCh38
rs775595291 Chr.15: 48878541-48878541 on Build GRCh38
rs979968640 Chr.15: 48878601-48878601 on Build GRCh38
rs748918804 Chr.15: 48878580-48878580 on Build GRCh38
rs777290972 Chr.15: 48878578-48878578 on Build GRCh38
rs764232155 Chr.15: 48878489-48878490 on Build GRCh38
rs539082813 Chr.15: 48878576-48878576 on Build GRCh38
rs910187141 Chr.15: 48878592-48878592 on Build GRCh38
rs759416600 Chr.15: 48878438-48878438 on Build GRCh38
rs758716473 Chr.15: 48878543-48878543 on Build GRCh38
rs753906069 Chr.15: 48878450-48878450 on Build GRCh38
rs758767752 Chr.15: 48878462-48878462 on Build GRCh38
rs751478270 Chr.15: 48878421-48878421 on Build GRCh38
rs759447313 Chr.15: 48878466-48878467 on Build GRCh38
rs371640324 Chr.15: 48878574-48878574 on Build GRCh38
rs772790872 Chr.15: 48878510-48878510 on Build GRCh38
rs759489672 Chr.15: 48878525-48878525 on Build GRCh38
rs765412776 Chr.15: 48878536-48878536 on Build GRCh38
rs767422505 Chr.15: 48878529-48878529 on Build GRCh38
rs774193104 Chr.15: 48878603-48878603 on Build GRCh38
rs781603165 Chr.15: 48878573-48878573 on Build GRCh38

View Full Product Details