Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 35577077-35577316 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKBP5
Entrez Gene ID
2289
Gene Name
FK506 binding protein 5
Gene Aliases
AIG6, FKBP51, FKBP54, P54, PPIase, Ptg-10
UniGene
Hs.407190
Gene Chromosome Location
Chr.6: 35573585-35728583 on Build GRCh38
Transcripts
NM_001145775.2 NM_001145776.1 NM_004117.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1754758 c.1057G>A p.G353S Chr.6: 35577203-35577203 on Build GRCh38
3921572 c.1034G>A p.G345E Chr.6: 35577226-35577226 on Build GRCh38
6664271 c.1179G>A p.M393I Chr.6: 35577081-35577081 on Build GRCh38
3921571 c.1068G>A p.R356R Chr.6: 35577192-35577192 on Build GRCh38
5870480 c.1028C>A p.A343D Chr.6: 35577232-35577232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755375739 Chr.6: 35577182-35577182 on Build GRCh38
rs34866878 Chr.6: 35577165-35577165 on Build GRCh38
rs779473333 Chr.6: 35577187-35577187 on Build GRCh38
rs375912702 Chr.6: 35577266-35577266 on Build GRCh38
rs753049321 Chr.6: 35577198-35577198 on Build GRCh38
rs767683901 Chr.6: 35577140-35577140 on Build GRCh38
rs547741786 Chr.6: 35577251-35577251 on Build GRCh38
rs140740382 Chr.6: 35577211-35577211 on Build GRCh38
rs868641354 Chr.6: 35577101-35577101 on Build GRCh38
rs370284375 Chr.6: 35577141-35577141 on Build GRCh38
rs760776253 Chr.6: 35577142-35577142 on Build GRCh38
rs200398524 Chr.6: 35577238-35577238 on Build GRCh38
rs139316938 Chr.6: 35577133-35577133 on Build GRCh38
rs143893856 Chr.6: 35577164-35577164 on Build GRCh38
rs775476348 Chr.6: 35577254-35577254 on Build GRCh38
rs199684698 Chr.6: 35577245-35577245 on Build GRCh38
rs759626624 Chr.6: 35577165-35577165 on Build GRCh38
rs146787424 Chr.6: 35577112-35577112 on Build GRCh38
rs762696275 Chr.6: 35577255-35577255 on Build GRCh38
rs916481623 Chr.6: 35577162-35577162 on Build GRCh38
rs769677713 Chr.6: 35577252-35577252 on Build GRCh38
rs371249911 Chr.6: 35577259-35577259 on Build GRCh38
rs374789777 Chr.6: 35577208-35577208 on Build GRCh38
rs1024217045 Chr.6: 35577131-35577131 on Build GRCh38
rs759488963 Chr.6: 35577106-35577106 on Build GRCh38
rs762796041 Chr.6: 35577138-35577138 on Build GRCh38
rs760831586 Chr.6: 35577267-35577267 on Build GRCh38
rs766400491 Chr.6: 35577160-35577160 on Build GRCh38
rs777596648 Chr.6: 35577210-35577210 on Build GRCh38
rs766453660 Chr.6: 35577269-35577269 on Build GRCh38

View Full Product Details