Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 117532177-117532448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCBLD1
Entrez Gene ID
285761
Gene Name
discoidin, CUB and LCCL domain containing 1
Gene Aliases
dJ94G16.1
UniGene
Hs.658304
Gene Chromosome Location
Chr.6: 117480346-117569858 on Build GRCh38
Transcripts
NM_173674.2 XM_005266941.1 XM_006715463.1 XM_011535771.1 XM_011535773.2 XM_011535774.2 XM_017010775.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6040592 c.719_719+3delAGTA p.? Chr.6: 117532393-117532396 on Build GRCh38
6450338 c.679C>G p.R227G Chr.6: 117532353-117532353 on Build GRCh38
483336 c.708T>C p.V236V Chr.6: 117532382-117532382 on Build GRCh38
3857827 c.600C>G p.C200W Chr.6: 117532274-117532274 on Build GRCh38
6640610 c.597G>A p.L199L Chr.6: 117532271-117532271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762578801 Chr.6: 117532400-117532401 on Build GRCh38
rs1032320941 Chr.6: 117532401-117532402 on Build GRCh38
rs757348762 Chr.6: 117532357-117532357 on Build GRCh38
rs748398337 Chr.6: 117532228-117532228 on Build GRCh38
rs375932606 Chr.6: 117532220-117532220 on Build GRCh38
rs996911300 Chr.6: 117532342-117532342 on Build GRCh38
rs192563915 Chr.6: 117532338-117532338 on Build GRCh38
rs374512343 Chr.6: 117532415-117532415 on Build GRCh38
rs769039254 Chr.6: 117532396-117532396 on Build GRCh38
rs779988320 Chr.6: 117532223-117532223 on Build GRCh38
rs772484448 Chr.6: 117532267-117532267 on Build GRCh38
rs750392207 Chr.6: 117532321-117532321 on Build GRCh38
rs201497239 Chr.6: 117532251-117532251 on Build GRCh38
rs766284839 Chr.6: 117532335-117532335 on Build GRCh38
rs748566955 Chr.6: 117532275-117532275 on Build GRCh38
rs867739877 Chr.6: 117532290-117532290 on Build GRCh38
rs139193542 Chr.6: 117532339-117532339 on Build GRCh38
rs768152858 Chr.6: 117532289-117532289 on Build GRCh38
rs749771493 Chr.6: 117532246-117532246 on Build GRCh38
rs777944366 Chr.6: 117532245-117532245 on Build GRCh38
rs373947907 Chr.6: 117532370-117532370 on Build GRCh38
rs76080980 Chr.6: 117532248-117532248 on Build GRCh38
rs754059942 Chr.6: 117532353-117532353 on Build GRCh38
rs894776327 Chr.6: 117532312-117532312 on Build GRCh38
rs764768860 Chr.6: 117532320-117532320 on Build GRCh38
rs761274265 Chr.6: 117532305-117532305 on Build GRCh38
rs747325556 Chr.6: 117532388-117532388 on Build GRCh38
rs1028414436 Chr.6: 117532344-117532344 on Build GRCh38
rs776183051 Chr.6: 117532397-117532397 on Build GRCh38
rs775772271 Chr.6: 117532271-117532271 on Build GRCh38
rs779038164 Chr.6: 117532361-117532361 on Build GRCh38
rs377575899 Chr.6: 117532384-117532384 on Build GRCh38
rs772785627 Chr.6: 117532416-117532416 on Build GRCh38
rs751364552 Chr.6: 117532337-117532337 on Build GRCh38
rs756340418 Chr.6: 117532237-117532237 on Build GRCh38
rs746371750 Chr.6: 117532259-117532259 on Build GRCh38
rs779241924 Chr.6: 117532266-117532266 on Build GRCh38
rs780427862 Chr.6: 117532387-117532387 on Build GRCh38
rs151207308 Chr.6: 117532278-117532278 on Build GRCh38
rs1046512413 Chr.6: 117532407-117532407 on Build GRCh38
rs761520742 Chr.6: 117532410-117532410 on Build GRCh38
rs772498841 Chr.6: 117532386-117532386 on Build GRCh38
rs77788962 Chr.6: 117532401-117532401 on Build GRCh38
rs776226463 Chr.6: 117532304-117532304 on Build GRCh38
rs762967204 Chr.6: 117532326-117532326 on Build GRCh38
rs763660240 Chr.6: 117532411-117532412 on Build GRCh38
rs752357664 Chr.6: 117532393-117532396 on Build GRCh38

View Full Product Details