Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54548174-54548431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL3L
Entrez Gene ID
54552
Gene Name
G protein nucleolar 3 like
Gene Aliases
GNL3B
UniGene
Hs.654677
Gene Chromosome Location
Chr.X: 54530211-54567287 on Build GRCh38
Transcripts
NM_001184819.1 NM_019067.5

SNP Detail

SNP ID Chromosome Location
rs765120166 Chr.X: 54548288-54548288 on Build GRCh38
rs371577497 Chr.X: 54548331-54548331 on Build GRCh38
rs758691564 Chr.X: 54548359-54548359 on Build GRCh38
rs5915137 Chr.X: 54548211-54548211 on Build GRCh38
rs200489809 Chr.X: 54548225-54548225 on Build GRCh38
rs773136749 Chr.X: 54548402-54548402 on Build GRCh38
rs769651264 Chr.X: 54548399-54548399 on Build GRCh38
rs753273204 Chr.X: 54548203-54548203 on Build GRCh38
rs776686747 Chr.X: 54548261-54548261 on Build GRCh38
rs749747958 Chr.X: 54548201-54548201 on Build GRCh38
rs138238719 Chr.X: 54548332-54548332 on Build GRCh38
rs1035007195 Chr.X: 54548246-54548246 on Build GRCh38
rs747673682 Chr.X: 54548216-54548216 on Build GRCh38
rs202012678 Chr.X: 54548215-54548216 on Build GRCh38
rs145095477 Chr.X: 54548358-54548358 on Build GRCh38
rs150423737 Chr.X: 54548312-54548312 on Build GRCh38
rs368637768 Chr.X: 54548215-54548215 on Build GRCh38
rs374949500 Chr.X: 54548394-54548394 on Build GRCh38
rs142870842 Chr.X: 54548344-54548344 on Build GRCh38
rs957684408 Chr.X: 54548326-54548326 on Build GRCh38
rs772860862 Chr.X: 54548300-54548300 on Build GRCh38
rs961275769 Chr.X: 54548251-54548251 on Build GRCh38
rs747092909 Chr.X: 54548236-54548236 on Build GRCh38
rs140471367 Chr.X: 54548233-54548233 on Build GRCh38
rs148427770 Chr.X: 54548398-54548398 on Build GRCh38
rs780264570 Chr.X: 54548360-54548360 on Build GRCh38
rs754596051 Chr.X: 54548343-54548343 on Build GRCh38
rs778439686 Chr.X: 54548195-54548195 on Build GRCh38
rs201402960 Chr.X: 54548385-54548385 on Build GRCh38
rs200669121 Chr.X: 54548232-54548232 on Build GRCh38
rs771691343 Chr.X: 54548407-54548410 on Build GRCh38
rs754411514 Chr.X: 54548347-54548347 on Build GRCh38
rs768662560 Chr.X: 54548254-54548254 on Build GRCh38

View Full Product Details