Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 30885563-30885836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COCH
Entrez Gene ID
1690
Gene Name
cochlin
Gene Aliases
COCH-5B2, COCH5B2, DFNA9
UniGene
Hs.21016
Gene Chromosome Location
Chr.14: 30874496-30890618 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 XM_017021073.1 NR_038356.1

Gene Symbol
LOC100506071
Entrez Gene ID
100506071
Gene Name
uncharacterized LOC100506071
Gene Aliases
-
UniGene
Hs.729195
Gene Chromosome Location
Chr.14: 30876179-30889808 on Build GRCh38
Transcripts
NM_001135058.1 NM_004086.2 XM_017021071.1 XM_017021072.1 XM_017021073.1 NR_038356.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3495646 c.929G>A p.G310E Chr.14: 30885589-30885589 on Build GRCh38
2034228 c.971G>A p.R324Q Chr.14: 30885806-30885806 on Build GRCh38
4775902 c.918_937del20 p.E307fs*6 Chr.14: 30885578-30885597 on Build GRCh38
7280553 c.931A>G p.M311V Chr.14: 30885591-30885591 on Build GRCh38
6632170 c.523C>T p.R175W Chr.14: 30885805-30885805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758676960 Chr.14: 30885768-30885768 on Build GRCh38
rs28362776 Chr.14: 30885761-30885761 on Build GRCh38
rs748731866 Chr.14: 30885806-30885806 on Build GRCh38
rs937599616 Chr.14: 30885623-30885623 on Build GRCh38
rs751416019 Chr.14: 30885633-30885633 on Build GRCh38
rs767389910 Chr.14: 30885635-30885635 on Build GRCh38
rs1046194994 Chr.14: 30885691-30885691 on Build GRCh38
rs764852134 Chr.14: 30885611-30885611 on Build GRCh38
rs772709865 Chr.14: 30885617-30885617 on Build GRCh38
rs776982710 Chr.14: 30885805-30885805 on Build GRCh38
rs149342451 Chr.14: 30885587-30885587 on Build GRCh38
rs760438334 Chr.14: 30885758-30885758 on Build GRCh38
rs35720988 Chr.14: 30885775-30885776 on Build GRCh38
rs1037985775 Chr.14: 30885640-30885640 on Build GRCh38
rs766266455 Chr.14: 30885628-30885628 on Build GRCh38
rs137925937 Chr.14: 30885653-30885653 on Build GRCh38
rs143509273 Chr.14: 30885773-30885773 on Build GRCh38
rs899301058 Chr.14: 30885683-30885683 on Build GRCh38
rs779113407 Chr.14: 30885769-30885769 on Build GRCh38
rs766663765 Chr.14: 30885790-30885791 on Build GRCh38
rs906626064 Chr.14: 30885756-30885756 on Build GRCh38
rs747378845 Chr.14: 30885791-30885791 on Build GRCh38
rs761251617 Chr.14: 30885597-30885597 on Build GRCh38
rs758491902 Chr.14: 30885792-30885792 on Build GRCh38
rs763064870 Chr.14: 30885786-30885786 on Build GRCh38
rs762762265 Chr.14: 30885620-30885620 on Build GRCh38
rs1012436139 Chr.14: 30885701-30885701 on Build GRCh38
rs756651532 Chr.14: 30885638-30885638 on Build GRCh38
rs886157814 Chr.14: 30885695-30885695 on Build GRCh38
rs779087171 Chr.14: 30885753-30885753 on Build GRCh38
rs757258736 Chr.14: 30885658-30885658 on Build GRCh38
rs753785426 Chr.14: 30885788-30885788 on Build GRCh38
rs368008430 Chr.14: 30885634-30885634 on Build GRCh38
rs1014408161 Chr.14: 30885684-30885684 on Build GRCh38
rs1045135091 Chr.14: 30885744-30885744 on Build GRCh38
rs879468608 Chr.14: 30885738-30885738 on Build GRCh38
rs769102282 Chr.14: 30885804-30885804 on Build GRCh38

View Full Product Details