Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67121220-67121463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM2A
Entrez Gene ID
22992
Gene Name
lysine demethylase 2A
Gene Aliases
CXXC8, FBL11, FBL7, FBXL11, JHDM1A, LILINA
UniGene
Hs.124147
Gene Chromosome Location
Chr.11: 67119210-67258082 on Build GRCh38
Transcripts
NM_012308.2 NR_027473.1 XM_011544860.2 XM_017017391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7157934 c.21G>A p.R7R Chr.11: 67121337-67121337 on Build GRCh38
931009 c.28T>C p.Y10H Chr.11: 67121344-67121344 on Build GRCh38
5627435 c.10G>A p.E4K Chr.11: 67121326-67121326 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779700263 Chr.11: 67121379-67121379 on Build GRCh38
rs533089864 Chr.11: 67121401-67121401 on Build GRCh38
rs1023767583 Chr.11: 67121305-67121305 on Build GRCh38
rs781683286 Chr.11: 67121377-67121378 on Build GRCh38
rs760718472 Chr.11: 67121272-67121272 on Build GRCh38
rs990957756 Chr.11: 67121260-67121260 on Build GRCh38
rs151044827 Chr.11: 67121325-67121325 on Build GRCh38
rs960917454 Chr.11: 67121252-67121252 on Build GRCh38
rs200263280 Chr.11: 67121316-67121316 on Build GRCh38
rs745508755 Chr.11: 67121370-67121370 on Build GRCh38
rs780727557 Chr.11: 67121365-67121365 on Build GRCh38
rs777629476 Chr.11: 67121334-67121334 on Build GRCh38
rs757032822 Chr.11: 67121362-67121362 on Build GRCh38
rs748753950 Chr.11: 67121385-67121385 on Build GRCh38
rs773347644 Chr.11: 67121403-67121403 on Build GRCh38
rs369716655 Chr.11: 67121397-67121397 on Build GRCh38
rs973881806 Chr.11: 67121377-67121377 on Build GRCh38
rs769293835 Chr.11: 67121388-67121388 on Build GRCh38
rs769498245 Chr.11: 67121373-67121373 on Build GRCh38
rs963199446 Chr.11: 67121345-67121345 on Build GRCh38
rs774882809 Chr.11: 67121389-67121389 on Build GRCh38
rs929649701 Chr.11: 67121410-67121410 on Build GRCh38
rs774024462 Chr.11: 67121421-67121421 on Build GRCh38
rs771556624 Chr.11: 67121371-67121372 on Build GRCh38
rs750611586 Chr.11: 67121268-67121268 on Build GRCh38
rs766414564 Chr.11: 67121289-67121289 on Build GRCh38
rs186824385 Chr.11: 67121339-67121339 on Build GRCh38

View Full Product Details