Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27360655-27360926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4R
Entrez Gene ID
3566
Gene Name
interleukin 4 receptor
Gene Aliases
CD124, IL-4RA, IL4RA
UniGene
Hs.513457
Gene Chromosome Location
Chr.16: 27313668-27364778 on Build GRCh38
Transcripts
NM_000418.3 NM_001257406.1 NM_001257407.1 NM_001257997.1 XM_005255308.2 XM_006721043.1 XM_011545825.1 XM_011545826.2 XM_011545827.2 XM_011545828.2 XM_011545829.1 XM_011545830.2 XM_011545831.1 XM_011545832.1 XM_011545833.2 XM_011545834.2 XM_017023211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4567809 c.882_883GG>TA p.Q294_E295>HK Chr.16: 27360798-27360799 on Build GRCh38
3782941 c.874C>T p.R292* Chr.16: 27360790-27360790 on Build GRCh38
3108732 c.869G>A p.R290Q Chr.16: 27360785-27360785 on Build GRCh38
3969485 c.899+1G>A p.? Chr.16: 27360816-27360816 on Build GRCh38
7185630 c.861G>A p.W287* Chr.16: 27360777-27360777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757851061 Chr.16: 27360773-27360773 on Build GRCh38
rs780518893 Chr.16: 27360896-27360896 on Build GRCh38
rs747723964 Chr.16: 27360905-27360905 on Build GRCh38
rs981287700 Chr.16: 27360901-27360901 on Build GRCh38
rs1032196613 Chr.16: 27360751-27360751 on Build GRCh38
rs372238631 Chr.16: 27360707-27360707 on Build GRCh38
rs1008353852 Chr.16: 27360736-27360736 on Build GRCh38
rs780758274 Chr.16: 27360781-27360781 on Build GRCh38
rs750727918 Chr.16: 27360844-27360844 on Build GRCh38
rs753969425 Chr.16: 27360815-27360815 on Build GRCh38
rs746501301 Chr.16: 27360778-27360778 on Build GRCh38
rs149570990 Chr.16: 27360780-27360780 on Build GRCh38
rs754690077 Chr.16: 27360893-27360893 on Build GRCh38
rs143309296 Chr.16: 27360791-27360791 on Build GRCh38
rs551180670 Chr.16: 27360757-27360757 on Build GRCh38
rs368332343 Chr.16: 27360705-27360705 on Build GRCh38
rs755154229 Chr.16: 27360871-27360871 on Build GRCh38
rs376794576 Chr.16: 27360718-27360718 on Build GRCh38
rs754374806 Chr.16: 27360728-27360728 on Build GRCh38
rs954481412 Chr.16: 27360881-27360881 on Build GRCh38
rs760743444 Chr.16: 27360793-27360793 on Build GRCh38
rs995930153 Chr.16: 27360841-27360841 on Build GRCh38
rs965795870 Chr.16: 27360753-27360753 on Build GRCh38
rs769790595 Chr.16: 27360785-27360785 on Build GRCh38
rs1055830674 Chr.16: 27360694-27360694 on Build GRCh38
rs995814340 Chr.16: 27360717-27360717 on Build GRCh38
rs747977051 Chr.16: 27360784-27360784 on Build GRCh38
rs751189046 Chr.16: 27360892-27360892 on Build GRCh38
rs898473541 Chr.16: 27360706-27360706 on Build GRCh38
rs887326901 Chr.16: 27360723-27360723 on Build GRCh38
rs967261813 Chr.16: 27360882-27360882 on Build GRCh38
rs987564540 Chr.16: 27360891-27360891 on Build GRCh38
rs746269184 Chr.16: 27360746-27360746 on Build GRCh38
rs779845396 Chr.16: 27360849-27360849 on Build GRCh38
rs764097618 Chr.16: 27360794-27360794 on Build GRCh38
rs34538949 Chr.16: 27360855-27360856 on Build GRCh38
rs1029191279 Chr.16: 27360855-27360855 on Build GRCh38
rs533865105 Chr.16: 27360790-27360790 on Build GRCh38
rs199515810 Chr.16: 27360819-27360819 on Build GRCh38
rs566962057 Chr.16: 27360870-27360870 on Build GRCh38
rs758889124 Chr.16: 27360847-27360847 on Build GRCh38
rs765809790 Chr.16: 27360825-27360825 on Build GRCh38
rs764572498 Chr.16: 27360719-27360719 on Build GRCh38

View Full Product Details