Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1949053-1949304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP1B
Entrez Gene ID
196513
Gene Name
decapping mRNA 1B
Gene Aliases
DCP1
UniGene
Hs.130934
Gene Chromosome Location
Chr.12: 1941591-2004535 on Build GRCh38
Transcripts
NM_152640.4 NR_135060.1 XM_011520927.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
692899 c.1633C>T p.L545F Chr.12: 1949226-1949226 on Build GRCh38
4872564 c.1323C>T p.S441S Chr.12: 1949230-1949230 on Build GRCh38
692898 c.1591G>T p.A531S Chr.12: 1949268-1949268 on Build GRCh38
4040716 c.1576T>C p.S526P Chr.12: 1949283-1949283 on Build GRCh38
226274 c.1715T>A p.I572N Chr.12: 1949144-1949144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200553703 Chr.12: 1949196-1949196 on Build GRCh38
rs753450583 Chr.12: 1949259-1949259 on Build GRCh38
rs765479804 Chr.12: 1949074-1949074 on Build GRCh38
rs11835821 Chr.12: 1949127-1949127 on Build GRCh38
rs773965946 Chr.12: 1949278-1949278 on Build GRCh38
rs769038532 Chr.12: 1949273-1949273 on Build GRCh38
rs769549804 Chr.12: 1949116-1949116 on Build GRCh38
rs549502503 Chr.12: 1949135-1949135 on Build GRCh38
rs771282481 Chr.12: 1949268-1949268 on Build GRCh38
rs753972795 Chr.12: 1949071-1949071 on Build GRCh38
rs560777764 Chr.12: 1949109-1949109 on Build GRCh38
rs140288898 Chr.12: 1949148-1949148 on Build GRCh38
rs759399707 Chr.12: 1949232-1949232 on Build GRCh38
rs775709702 Chr.12: 1949191-1949191 on Build GRCh38
rs770679389 Chr.12: 1949177-1949177 on Build GRCh38
rs747662585 Chr.12: 1949211-1949211 on Build GRCh38
rs147171506 Chr.12: 1949159-1949159 on Build GRCh38
rs751290770 Chr.12: 1949231-1949231 on Build GRCh38
rs956623680 Chr.12: 1949175-1949175 on Build GRCh38
rs1026662732 Chr.12: 1949102-1949102 on Build GRCh38
rs758143883 Chr.12: 1949154-1949154 on Build GRCh38
rs770535391 Chr.12: 1949221-1949222 on Build GRCh38
rs142458598 Chr.12: 1949276-1949276 on Build GRCh38
rs200121287 Chr.12: 1949217-1949217 on Build GRCh38
rs778673866 Chr.12: 1949269-1949269 on Build GRCh38
rs563243871 Chr.12: 1949140-1949140 on Build GRCh38
rs748667140 Chr.12: 1949282-1949282 on Build GRCh38
rs747504838 Chr.12: 1949253-1949253 on Build GRCh38
rs780001147 Chr.12: 1949081-1949081 on Build GRCh38
rs2240612 Chr.12: 1949079-1949079 on Build GRCh38
rs902119422 Chr.12: 1949251-1949251 on Build GRCh38
rs746198934 Chr.12: 1949125-1949125 on Build GRCh38
rs534343216 Chr.12: 1949176-1949176 on Build GRCh38
rs796559700 Chr.12: 1949087-1949087 on Build GRCh38
rs755719334 Chr.12: 1949173-1949173 on Build GRCh38
rs1033313086 Chr.12: 1949086-1949086 on Build GRCh38
rs747702830 Chr.12: 1949111-1949111 on Build GRCh38
rs559902808 Chr.12: 1949073-1949073 on Build GRCh38
rs138632437 Chr.12: 1949167-1949167 on Build GRCh38
rs149253407 Chr.12: 1949230-1949230 on Build GRCh38
rs747285683 Chr.12: 1949272-1949272 on Build GRCh38
rs376130978 Chr.12: 1949091-1949091 on Build GRCh38
rs1025281210 Chr.12: 1949152-1949152 on Build GRCh38
rs146350416 Chr.12: 1949277-1949277 on Build GRCh38
rs146996705 Chr.12: 1949149-1949149 on Build GRCh38
rs773023576 Chr.12: 1949119-1949119 on Build GRCh38
rs781329577 Chr.12: 1949166-1949166 on Build GRCh38
rs761839403 Chr.12: 1949207-1949207 on Build GRCh38
rs758421658 Chr.12: 1949077-1949077 on Build GRCh38
rs767525433 Chr.12: 1949248-1949248 on Build GRCh38
rs144383893 Chr.12: 1949249-1949249 on Build GRCh38
rs372610087 Chr.12: 1949076-1949076 on Build GRCh38
rs16928961 Chr.12: 1949158-1949158 on Build GRCh38
rs138120926 Chr.12: 1949153-1949153 on Build GRCh38
rs776011833 Chr.12: 1949130-1949130 on Build GRCh38
rs777141858 Chr.12: 1949199-1949199 on Build GRCh38
rs773364842 Chr.12: 1949229-1949229 on Build GRCh38
rs777412923 Chr.12: 1949254-1949254 on Build GRCh38

View Full Product Details