Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 16597381-16597628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAZL
Entrez Gene ID
1618
Gene Name
deleted in azoospermia like
Gene Aliases
DAZH, DAZL1, DAZLA, SPGYLA
UniGene
Hs.131179
Gene Chromosome Location
Chr.3: 16586792-16605499 on Build GRCh38
Transcripts
NM_001190811.1 NM_001351.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1203016 c.243-1G>A p.? Chr.3: 16597542-16597542 on Build GRCh38
6291577 c.293A>G p.E98G Chr.3: 16597491-16597491 on Build GRCh38
4945156 c.284A>G p.K95R Chr.3: 16597500-16597500 on Build GRCh38
6215803 c.280C>A p.Q94K Chr.3: 16597504-16597504 on Build GRCh38
4541895 c.248G>A p.G83E Chr.3: 16597536-16597536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74208885 Chr.3: 16597456-16597456 on Build GRCh38
rs765151404 Chr.3: 16597549-16597549 on Build GRCh38
rs757052717 Chr.3: 16597416-16597416 on Build GRCh38
rs775863559 Chr.3: 16597493-16597493 on Build GRCh38
rs781084309 Chr.3: 16597425-16597425 on Build GRCh38
rs773178905 Chr.3: 16597550-16597550 on Build GRCh38
rs771384044 Chr.3: 16597460-16597460 on Build GRCh38
rs373680387 Chr.3: 16597415-16597415 on Build GRCh38
rs772339113 Chr.3: 16597474-16597474 on Build GRCh38
rs367746828 Chr.3: 16597454-16597454 on Build GRCh38
rs748444130 Chr.3: 16597446-16597446 on Build GRCh38
rs745996141 Chr.3: 16597465-16597465 on Build GRCh38
rs376647546 Chr.3: 16597452-16597452 on Build GRCh38
rs780774408 Chr.3: 16597444-16597444 on Build GRCh38
rs756483007 Chr.3: 16597448-16597448 on Build GRCh38
rs748632383 Chr.3: 16597448-16597449 on Build GRCh38
rs569663997 Chr.3: 16597576-16597576 on Build GRCh38
rs759030609 Chr.3: 16597556-16597556 on Build GRCh38
rs372324455 Chr.3: 16597439-16597439 on Build GRCh38
rs867271446 Chr.3: 16597492-16597492 on Build GRCh38
rs200341557 Chr.3: 16597516-16597516 on Build GRCh38
rs992242205 Chr.3: 16597434-16597434 on Build GRCh38
rs762922974 Chr.3: 16597575-16597575 on Build GRCh38
rs773957250 Chr.3: 16597552-16597553 on Build GRCh38
rs770221939 Chr.3: 16597450-16597453 on Build GRCh38
rs766763008 Chr.3: 16597476-16597480 on Build GRCh38
rs751073695 Chr.3: 16597579-16597579 on Build GRCh38
rs767220381 Chr.3: 16597559-16597560 on Build GRCh38
rs761068673 Chr.3: 16597463-16597464 on Build GRCh38
rs761648553 Chr.3: 16597500-16597500 on Build GRCh38
rs773608954 Chr.3: 16597453-16597456 on Build GRCh38
rs75931701 Chr.3: 16597518-16597518 on Build GRCh38
rs866917727 Chr.3: 16597491-16597491 on Build GRCh38
rs754142452 Chr.3: 16597426-16597426 on Build GRCh38
rs774470685 Chr.3: 16597464-16597464 on Build GRCh38

View Full Product Details